Skip to Content
Merck
All Photos(1)

Key Documents

SRP4647

Sigma-Aldrich

Galectin-7 human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)

Synonym(s):

Gal-7, Galactein 7, Galectin7, HKL-14, PI7, p53-induced gene 1 protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

mol wt

~15.4 kDa

packaging

pkg of 50 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

endotoxin, tested

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... LGALS7(3963)

General description

LGALS7 (galectin-7) is from the galectin family of β-galactoside-binding proteins. It is mainly present in the stratified epithelia, hair follicles and thymic Hassall′s corpuscles. The protein localizes in the nucleus and cytoplasm, and exists as a homodimer. Recombinant Human Galectin-7 is a 15.4kDa protein consisting of 136 amino acid residues

Biochem/physiol Actions

LGALS7 (galectin-7) helps in maintaining skin homeostasis, particularly after skin injury. It is also involved in keratinocyte proliferation and differentiation via JNK1 (c-Jun N-terminal kinase)-miR-203-p63 pathway. LGALS7 is a biomarker for skin condition and skin barrier disruption. In HeLa cells, it induces apoptosis most likely via JNK signaling. LGALS7 is downregulated in SV40-transformed cells and squamous cell carcinoma cells. Change in its activity/expression is associated with the progression of various epithelia-associated cancers. LGALS7 also helps in postmenstrual endometrial re-epithelialization.

Physical form

Sterile filtered and Lyophilized without additives.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute in water to a concentration of 0.1-1.0 mg/mL. This solution can then be diluted into other aqueous buffers.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Galectin-7 Regulates Keratinocyte Proliferation and Differentiation through JNK-miR-203-p63 Signaling.
Chen HL, et al.
The Journal of Investigative Dermatology, 136, 182-182 (2016)
Galectin-7 is important for normal uterine repair following menstruation.
Evans J, et al.
Molecular Human Reproduction, 20, 787-787 (2014)
Galectin-7 in the stratum corneum: a biomarker of the skin barrier function.
Niiyama S, et al.
International Journal of Cosmetic Science, 38, 487-487 (2016)
A Mutation in the Carbohydrate Recognition Domain Drives a Phenotypic Switch in the Role of Galectin-7 in Prostate Cancer.
Labrie M, et al.
PLoS ONE, 0, e0131307-e0131307 (2015)
Jemma Evans et al.
Molecular human reproduction, 20(8), 787-798 (2014-05-02)
Menstruation involves the shedding of the functional layer of the endometrium in the absence of pregnancy. At sites where tissue shedding is complete, re-epithelialization of the tissue is essential for repair and termination of bleeding. The complement of growth factors

Articles

Find the right lectin for your research with our lectin selection guide, organized by lectin source/species, carbohydrate specificity, blood group specificity, and more.

Find the right lectin for your research with our lectin selection guide, organized by lectin source/species, carbohydrate specificity, blood group specificity, and more.

Find the right lectin for your research with our lectin selection guide, organized by lectin source/species, carbohydrate specificity, blood group specificity, and more.

Find the right lectin for your research with our lectin selection guide, organized by lectin source/species, carbohydrate specificity, blood group specificity, and more.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service