Skip to Content
Merck
All Photos(1)

Key Documents

SRP2083

Sigma-Aldrich

WT-1 (-KTS) human

recombinant, expressed in insect cells, ≥60% (SDS-PAGE)

Synonym(s):

AWT1, GUD, NPHS4, WAGR, WIT-2, WT33

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.77

biological source

human

recombinant

expressed in insect cells

Assay

≥60% (SDS-PAGE)

form

frozen liquid

mol wt

~55.6 kDa

packaging

pkg of 5 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

250 μg/mL

color

clear colorless

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... WT1(7490)

General description

Wilms tumor 1 (WT1) is encoded by the gene mapped to human chromosome 11p13. The encoded protein is expressed at high levels in nephrons, but at low levels in gonads, visceral adipose tissue, mesothelium and bone marrow cells. In addition, it is also abundantly expressed in renal podocytes in adults and in parietal epithelial cells during early days.

Biochem/physiol Actions

WT-1, the product of Wilms′ tumor suppressor gene Wt1, is a nuclear protein with structural motifs characteristic of transcription factors, including four C-terminal zinc fingers. While different pre-mRNA processing could result in 16 isoforms of the protein, inclusion or exclusion of exon 5 and the three amino acids (KTS) between zinc fingers 3 and 4 largely affects the activity of WT1 protein. Such a complex post-transcriptional regulation, particularly in splicing, may represent a major regulatory mechanism for tumorigenesis of the Wilms′ tumor. WT1 (-KTS) appears to have different binding affinity to both DNA and RNA compared to the +KTS form.
Wilms tumor 1 (WT1) is involved in various vertebrate developmental processes such as cell differentiation and proliferation, apoptosis and regulation of epithelial/mesenchymal characteristics. It serves as a tumor suppressor gene but might also possess oncogenic property. The encoded protein regulates mRNA splicing and interactions between protein. It is associated with cancers and as well as kidney function. Mutations in the gene leads to Denys-Drash syndrome, resulting in renal and genitourinary tract abnormalities. Deletion in the germline WT1 gene causes Wilms′ tumor. This gene is highly expressed in a various types of cancers.

Physical form

Clear and colorless frozen liquid solution

Preparation Note

Use a manual defrost freezer and avoid repeated freeze-thaw cycles. While working, please keep sample on ice.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D A Haber et al.
Science (New York, N.Y.), 262(5142), 2057-2059 (1993-12-24)
A human Wilms tumor cell line (RM1) was developed to test the tumor suppressor activity of WT1, a zinc finger transcription factor that is expressed in the developing human kidney and is mutationally inactivated in a subset of Wilms tumors.
The Role of WT1 in Embryonic Development and Normal Organ Homeostasis.
Wilm B and Mu?oz-Chapuli R
Methods in Molecular Biology, 1467, 23-39 (2016)
Clinical Aspects of WT1 and the Kidney.
Miller-Hodges E, et al.
Methods in Molecular Biology, 1467, 15-21 (2016)
Complete sequencing of the Fugu WAGR region from WT1 to PAX6: Dramatic compaction and conservation of synteny with human chromosome 11p13
Colin M, et al.
Proceedings of the National Academy of Sciences of the USA, 95(22), 13068-13072 (1998)
J A Kreidberg et al.
Cell, 74(4), 679-691 (1993-08-27)
In humans, germline mutations of the WT-1 tumor suppressor gene are associated with both Wilms' tumors and urogenital malformations. To develop a model system for the molecular analysis of urogenital development, we introduced a mutation into the murine WT-1 tumor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service