Skip to Content
Merck
All Photos(2)

Key Documents

SML3904

Sigma-Aldrich

FerroLOXIN-1

≥96% (HPLC)

Synonym(s):

N-(3-Fluoro-4-(trifluoromethyl)benzyl)-1-(3-fluorophenyl)-4-phenyl-1H-imidazol-2-amine, N-[[3-Fluoro-4-(trifluoromethyl)phenyl]methyl]-1-(3-fluorophenyl)-4-phenyl-1H-imidazol-2-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H16F5N3
CAS Number:
Molecular Weight:
429.39
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥96% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

shipped in

wet ice

storage temp.

-10 to -25°C

Biochem/physiol Actions

15LOX/PEBP1 catalytic complex inhibitor that prevents cell death by ferroptosis, but not necroptosis, pyroptosis, or apoptosis.


FerroLOXIN-1 is a ferroptosis inhibitor that selectively targets the 15LOX/PEBP1 catalytic complex without affecting the biosynthesis of pro-/anti-inflammatory lipid mediators. FerroLOXIN-1 prevents 0.5 μM GPX4 inhibitor RSL3-induced ferroptosis in HBE cell cultures (IC50 = 84 nM post 20h RSL3 treatment) and rescues mice from total body irradiation (TBI)-induced death in vivo (Cpd administered via 25 mg/kg ip. 20h post 9.25Gy irradiation; survival on day 27 = 75% vs. 0% on day 19 of untreated control). FerroLOXIN-1 does not exert its efficacy via radical scavenging or iron-chelating activity, nor does it prevent cell death by necroptosis, pyroptosis, or apoptosis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Targeting lipoxygenases to suppress ferroptotic cell death
Proceedings of the National Academy of Sciences of the USA, 120(29) (2023)
Discovering selective antiferroptotic inhibitors of the 15LOX/PEBP1 complex noninterfering with biosynthesis of lipid mediators
Proceedings of the National Academy of Sciences of the USA, 120(25) (2023)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service