Skip to Content
Merck
All Photos(1)

Documents

SML3593

Sigma-Aldrich

Hexachlorophene

≥95% (HPLC)

Synonym(s):

2,2′,3,3′,5,5′-Hexachloro-6,6′-dihydroxydiphenylmethane, 2,2′-Dihydroxy-3,3′,5,5′,6,6′-hexachlorodiphenylmethane, 2,2′-Methylenebis[3,4,6-trichlorophenol], Nabac

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H6Cl6O2
CAS Number:
Molecular Weight:
406.90
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.04

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

ClC1=C(C(O)=C(C=C1Cl)Cl)CC2=C(C(Cl)=CC(Cl)=C2O)Cl

Biochem/physiol Actions

Hexachlorophene is selective and potent N-acyl-phosphatidylethanolamine phospholipase D (NAPE-PLD) inhibitor. Hexachlorophene potently inhibits NAPE-PLD activity in cultured HEK293 cells. Hexachlorophene is an antibacterial compound used topically by health-care personnel to control cutaneous gram-positive infections. Also it appears that hexachlorophene is an inhibitor of virus entry for Severe fever with thrombocytopenia syndrome virus (SFTSV).

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Symmetrically substituted dichlorophenes inhibit N-acyl-phosphatidylethanolamine phospholipase D
The Journal of Biological Chemistry, 295(21), 7289-7300 (2020)
Screening of an FDA-Approved Drug Library with a Two-Tier System Identifies an Entry Inhibitor of Severe Fever with Thrombocytopenia Syndrome Virus
Viruses, 11, 385-385 (2019)
Identification of repositionable drugs with novel antimycotic activity by screening the Prestwick Chemical Library against emerging invasive moulds
Journal of global antimicrobial resistance, 21, 314-317 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service