Skip to Content
Merck
All Photos(1)

Key Documents

SML3360

Sigma-Aldrich

STM2457

≥98% (HPLC)

Synonym(s):

N-[(6-{[(Cyclohexylmethyl)amino]methyl}imidazo[1,2-a]pyridin-2-yl)methyl]-4-oxo-4H-pyrido[1,2-a]pyrimidine-2-carboxamide, STM 2457, STM-2457

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H28N6O2
CAS Number:
Molecular Weight:
444.53
UNSPSC Code:
12352200
NACRES:
NA.21

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C1C=C(C(NCC2=CN3C=C(CNCC4CCCCC4)C=CC3=N2)=O)N=C5C=CC=CN15

InChI

1S/C25H28N6O2/c32-24-12-21(29-23-8-4-5-11-31(23)24)25(33)27-15-20-17-30-16-19(9-10-22(30)28-20)14-26-13-18-6-2-1-3-7-18/h4-5,8-12,16-18,26H,1-3,6-7,13-15H2,(H,27,33)

InChI key

OBERVORNENYOLE-UHFFFAOYSA-N

Biochem/physiol Actions

Potent and highly selective SAM-binding site METTL3 (MT-A70) m6A methyltransferase inhibitor with anti-AML efficacy in cultures and in vivo.
STM2457 is a potent and highly selective SAM-binding site METTL3 (MT-A70) m6A methyltransferase inhibitor (IC50 = 16.9 nM, Kd = 1.4 nM by SPR, using METTL3-METTL14 complex; no potency toward other RNA/DNA/protein methyltransferases or 486 kinases). STM2457 selectively inhibits human AML proliferation (IC50 = 0.6-10.3 µM) and the clonogenic potential of primary mouse AML, but not normal mouse haematopoietic stem/progenitor or human cord blood CD34+ cells. STM2457 impairs the expansion of AML patient-derived xenografts in mice in vivo (50 mg/kg i.p.) and significantly prolongs the animal′s lifespan. STM2457 is also reported to restrict β-coronavirus replication and spread.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nadine Körtel et al.
Nucleic acids research, 49(16), e92-e92 (2021-06-23)
N6-methyladenosine (m6A) is the most abundant internal RNA modification in eukaryotic mRNAs and influences many aspects of RNA processing. miCLIP (m6A individual-nucleotide resolution UV crosslinking and immunoprecipitation) is an antibody-based approach to map m6A sites with single-nucleotide resolution. However, due
Eliza Yankova et al.
Nature, 593(7860), 597-601 (2021-04-27)
N6-methyladenosine (m6A) is an abundant internal RNA modification1,2 that is catalysed predominantly by the METTL3-METTL14 methyltransferase complex3,4. The m6A methyltransferase METTL3 has been linked to the initiation and maintenance of acute myeloid leukaemia (AML), but the potential of therapeutic applications
Hannah M Burgess et al.
Genes & development, 35(13-14), 1005-1019 (2021-06-26)
N6-methyladenosine (m6A) is an abundant internal RNA modification, influencing transcript fate and function in uninfected and virus-infected cells. Installation of m6A by the nuclear RNA methyltransferase METTL3 occurs cotranscriptionally; however, the genomes of some cytoplasmic RNA viruses are also m6A-modified.
Qisheng Zuo et al.
EMBO reports, 23(1), e52491-e52491 (2021-11-09)
The unique characteristics of chicken primordial germ cells (PGCs) provide potential strategies for transgenic animal generation; however, insufficient PGC availability has limited their application. Regulation of bone morphogenic protein 4 (BMP4), a crucial factor for PGCs formation, may provide new

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service