Skip to Content
Merck
All Photos(1)

Key Documents

SML2377

Sigma-Aldrich

GSK′962

≥98% (HPLC)

Synonym(s):

1-[(5R)-4,5-Dihydro-5-phenyl-1H-pyrazol-1-yl]-2,2-dimethyl-1-propanone; 2,2-Dimethyl-1-(5(R)-phenyl-4,5-dihydro-pyrazol-1-yl)-propan-1-one, GSK 962, GSK 963 inactive control, GSK 963 negative control, GSK′ 962A, GSK′962A, GSK′963 inactive control, GSK′963 negative control, GSK-962, GSK-962A, GSK962, GSK962A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H18N2O
CAS Number:
Molecular Weight:
230.31
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(C(C)(C)C)N1[C@@H](C2=CC=CC=C2)CC=N1

Biochem/physiol Actions

GSK′962 (GSK′962A) corresponds to the inactive (R)-enantiomer of the highly potent and selective RIPK1 (RIP1) inhibitor GSK′963 (GSK′963A). GSK′962 serves as an ideal GSK′963 negative control compound for probing RIPK1-mediated responses both in vitro and in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenjing Liu et al.
Proceedings of the National Academy of Sciences of the United States of America, 115(7), E1475-E1484 (2018-02-01)
Tubular cell necrosis is a key histological feature of acute kidney injury (AKI). Necroptosis is a type of programed necrosis, which is executed by mixed lineage kinase domain-like protein (MLKL) upon its binding to the plasma membrane. Emerging evidence indicates
Mallary C Greenlee-Wacker et al.
Blood, 129(24), 3237-3244 (2017-05-06)
Community-associated methicillin-resistant Staphylococcus aureus (CA-MRSA) causes infections associated with extensive tissue damage and necrosis. In vitro, human neutrophils fed CA-MRSA lyse by an unknown mechanism that is inhibited by necrostatin-1, an allosteric inhibitor of receptor-interacting serine/threonine kinase 1 (RIPK-1). RIPK-1
Nivea Farias Luz et al.
Journal of immunology (Baltimore, Md. : 1950), 196(12), 5056-5063 (2016-05-18)
Leishmaniasis is an important parasitic disease found in the tropics and subtropics. Cutaneous and visceral leishmaniasis affect an estimated 1.5 million people worldwide. Despite its human health relevance, relatively little is known about the cell death pathways that control Leishmania
Dan Weng et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(20), 7391-7396 (2014-05-07)
A number of pathogens cause host cell death upon infection, and Yersinia pestis, infamous for its role in large pandemics such as the "Black Death" in medieval Europe, induces considerable cytotoxicity. The rapid killing of macrophages induced by Y. pestis
S B Berger et al.
Cell death discovery, 1, 15009-15009 (2015-01-01)
Necroptosis and signaling regulated by RIP1 kinase activity is emerging as a key driver of inflammation in a variety of disease settings. A significant amount has been learned about how RIP1 regulates necrotic cell death through the use of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service