Skip to Content
Merck
All Photos(1)

Documents

SML2023

Sigma-Aldrich

UCSF924NC

≥98% (HPLC)

Synonym(s):

6-Methyl-2-((3-(pyridin-4-yl)propylamino)methyl)quinolin-4(1H)-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H21N3O
CAS Number:
Molecular Weight:
307.39
UNSPSC Code:
51111800
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

CC1=CC2=C(NC(CNCCCC3=CC=NC=C3)=CC2=O)C=C1

Biochem/physiol Actions

UCSF924NC (Compound 9-6-16; ZINC000091707446) is the recommended negative control compound for the high-affinity dopamine D4 receptor (DRD4) partial agonist UCSF924 (Compound 9-6-24; ZINC000091726127). UCSF924NC exhibits a 1/2500-fold reduced D4 affinity when compared with that of UCSF924.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sheng Wang et al.
Science (New York, N.Y.), 358(6361), 381-386 (2017-10-21)
Dopamine receptors are implicated in the pathogenesis and treatment of nearly every neuropsychiatric disorder. Although thousands of drugs interact with these receptors, our molecular understanding of dopaminergic drug selectivity and design remains clouded. To illuminate dopamine receptor structure, function, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service