Skip to Content
Merck
All Photos(1)

Key Documents

SML1393

Sigma-Aldrich

ACT-462206

≥98% (HPLC)

Synonym(s):

(2S)-N-(3,5-Dimethylphenyl)-1-[(4-methoxyphenyl)sulfonyl]-2-pyrrolidinecarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24N2O4S
CAS Number:
Molecular Weight:
388.48
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

O=S(N1CCC[C@H]1C(NC2=CC(C)=CC(C)=C2)=O)(C3=CC=C(OC)C=C3)=O

InChI

1S/C20H24N2O4S/c1-14-11-15(2)13-16(12-14)21-20(23)19-5-4-10-22(19)27(24,25)18-8-6-17(26-3)7-9-18/h6-9,11-13,19H,4-5,10H2,1-3H3,(H,21,23)/t19-/m0/s1

InChI key

NHPQGZOBHSVTAQ-IBGZPJMESA-N

Biochem/physiol Actions

ACT-462206 is a orally active, brain penetrant, potent and selective dual orexin1/2 receptor antagonist (DORA). ACT-462206 decreases wakefulness, decreases sleep latency, and increases sleep efficacy in rats and dogs.
ACT-462206 is a proline sulphonamide with the potential to treat insomnia. On absorption, its displays asthenia, blurred vision, and fatigue as adverse events.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christoph Boss et al.
ChemMedChem, 9(11), 2486-2496 (2014-08-26)
The orexin system consists of two G-protein-coupled receptors, the orexin 1 and orexin 2 receptors, widely expressed in diverse regions of the brain, and two peptide agonists, orexin A and orexin B, which are produced in a small assembly of
Matthias Hoch et al.
Journal of clinical pharmacology, 54(9), 979-986 (2014-04-03)
A double-blind, placebo- and active comparator-controlled, randomized, single-ascending-dose study was conducted to investigate the safety, pharmacokinetics, and pharmacodynamics of ACT-462206, a novel dual orexin receptor antagonist. Healthy male subjects received single oral doses of 5, 25, 100, 200, 400, 1,000

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service