Skip to Content
Merck
All Photos(1)

Key Documents

SML0951

Sigma-Aldrich

Salubrinal

≥98% (HPLC), powder, eIF2α dephosphorylation inhibitor

Synonym(s):

(2E)-3-Phenyl-N-[2,2,2-trichloro-1-[[(8-quinolinylamino)thioxomethyl]amino]ethyl] 2-propenamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H17Cl3N4OS
CAS Number:
Molecular Weight:
479.81
UNSPSC Code:
12352200
NACRES:
NA.77

Product Name

Salubrinal, ≥98% (HPLC)

Quality Level

Assay

≥98% (HPLC)

form

powder

color

, White to brown to grey

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

S=C(NC(NC(=O)\C=C\c3ccccc3)C(Cl)(Cl)Cl)Nc1c2ncccc2ccc1

InChI

1S/C21H17Cl3N4OS/c22-21(23,24)19(27-17(29)12-11-14-6-2-1-3-7-14)28-20(30)26-16-10-4-8-15-9-5-13-25-18(15)16/h1-13,19H,(H,27,29)(H2,26,28,30)/b12-11+

InChI key

LCOIAYJMPKXARU-VAWYXSNFSA-N

Application

Salubrinal has been used to study the effect of stress-protective genes and pathways on organism development and lifespan. It has also been used to study the therapeutic potential of salubrinal in methamphetamine induced blood–brain barrier disruption.

Biochem/physiol Actions

Salubrinal is a selective Inhibitor of eIF2α (eukaryotic translation initiation factor 2 α-subunit) dephosphorylation, inhibiting global protein translation. Salubrinal is a specific inhibitor of ER Stress induced apoptosis.
Salubrinal is a selective Inhibitor of eIF2α dephosphorylation; specific inhibitor of ER Stress induced apoptosis.
Salubrinal maintains protein phosphorylation in order to offer protection against from the adverse effects of endoplasmic reticulum stress. Salubrinal prevents collagenase-mediated articular cartilage damage promoting downregulation of matrix metalloproteinase 13.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Muhammad M Hossain et al.
Toxicological sciences : an official journal of the Society of Toxicology, 167(1), 249-257 (2018-09-25)
Endoplasmic reticulum (ER) stress is a significant contributor to neurodegeneration and cognitive dysfunction. Recently, we reported that repeated exposure to the pyrethroid insecticide deltamethrin caused ER stress in the hippocampus of adult mice, which was accompanied by deficits in learning
K Hamamura et al.
Osteoarthritis and cartilage, 21(5), 764-772 (2013-03-12)
Stress to the endoplasmic reticulum (ER) and inflammatory cytokines induce expression and activity of matrix metalloproteinase 13 (MMP13). Since a synthetic agent, salubrinal, is known to alleviate ER stress and attenuate nuclear factor kappa B (NFκB) signaling, we addressed a
Rescue of a developmental arrest caused by a C. elegans heat-shock transcription-factor mutation by loss of ribosomal S6-kinase activity.
Chisnell P J D, et al.
bioRxiv, 310086-310086 (2018)
Protection of Bcl-2 by salubrinal.
Kessel D.
Biochemical and Biophysical Research Communications, 346(4), 1320-1323 (2006)
Shunsuke Sakai et al.
Journal of cellular physiology, 237(1), 1087-1099 (2021-10-15)
In the postpartum period, cows experience the uterine bacterial infection and develop the endometritis. To eliminate bacteria and recover from endometritis, endometrial epithelial and stromal cells secrete the cytokine and chemokine, such as interleukin 6 (IL-6), IL-8, and monocyte chemotactic

Articles

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service