Skip to Content
Merck
All Photos(1)

Documents

SML0636

Sigma-Aldrich

SR1664

≥98% (HPLC)

Synonym(s):

4′-[[2,3-Dimethyl-5-[[[(1S)-1-(4-nitrophenyl)ethyl]amino]carbonyl]-1H-indol-1-yl]methyl]- [1,1′-Biphenyl]-2-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H29N3O5
CAS Number:
Molecular Weight:
547.60
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

white to beige

solubility

DMSO: 15 mg/mL, clear

storage temp.

−20°C

InChI

1S/C33H29N3O5/c1-20-22(3)35(19-23-8-10-25(11-9-23)28-6-4-5-7-29(28)33(38)39)31-17-14-26(18-30(20)31)32(37)34-21(2)24-12-15-27(16-13-24)36(40)41/h4-18,21H,19H2,1-3H3,(H,34,37)(H,38,39)/t21-/m0/s1

InChI key

IIJDFXNUWZTHIM-NRFANRHFSA-N

Biochem/physiol Actions

SR1664 is a non-agonist PPARγ ligand and an inhibitor of Cdk5-mediated PPARγ phosphorylation. It has strong antidiabetic activity in two murine models of diabetes without the side effects normally asociated with the thiazolidinedione (TZD) antidiabetic PPARγ agonists. The TZD antidiabetics such as rosiglitazone and pioglitazone are full PPARγ agonists, but recent data have suggested that their separate activity, inhibition of the the obesity-linked PPARγ phosphorylation by Cdk5, is likely the more important activity for antidiabetic action. SR1664 has an IC50 of 80 nM in a competitive binding assay, blocked the Cdk5-mediated phosphorylation of PPARγ in vitro with IC50 between 20 and 200 nM and exhibited no PPARγ agonist activity.

Features and Benefits

This compound is featured on the CDKs page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Other Notes

Light sensitive

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Review properties, activators and inhibitors, and available products for researching cyclin-dependent kinases (CDKs).

Review properties, activators and inhibitors, and available products for researching cyclin-dependent kinases (CDKs).

Review properties, activators and inhibitors, and available products for researching cyclin-dependent kinases (CDKs).

Review properties, activators and inhibitors, and available products for researching cyclin-dependent kinases (CDKs).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service