Skip to Content
Merck
All Photos(1)

Key Documents

SML0345

Sigma-Aldrich

A 887826

≥98% (HPLC)

Synonym(s):

5-(4-butoxy-3-chlorophenyl)-N-[[2-(4-morpholinyl)-3-pyridinyl]methyl]-3-pyridinecarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H29ClN4O3
CAS Number:
Molecular Weight:
480.99
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: ≥5 mg/mL

storage temp.

2-8°C

SMILES string

CCCCOc1ccc(cc1Cl)-c2cncc(c2)C(=O)NCc3cccnc3N4CCOCC4

InChI

1S/C26H29ClN4O3/c1-2-3-11-34-24-7-6-19(15-23(24)27)21-14-22(17-28-16-21)26(32)30-18-20-5-4-8-29-25(20)31-9-12-33-13-10-31/h4-8,14-17H,2-3,9-13,18H2,1H3,(H,30,32)

InChI key

JPJGFWKHSMUKFO-UHFFFAOYSA-N

Biochem/physiol Actions

A-887826 is Nav1.8 channel blocker (IC50 = 11 nM) that blocks TTX-resistant sodium currents in rat dorsal root ganglion in a voltage dependent fashion. Oral administration of A-887826 to rats significantly attenuated tactile allodynia in a neuropathic pain model.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Voltage-gated sodium channels are present in most excitable cell membranes and play an important role in generating action potentials.

Voltage-gated sodium channels are present in most excitable cell membranes and play an important role in generating action potentials.

Voltage-gated sodium channels are present in most excitable cell membranes and play an important role in generating action potentials.

Voltage-gated sodium channels are present in most excitable cell membranes and play an important role in generating action potentials.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service