Skip to Content
Merck
All Photos(1)

Key Documents

SMB00436

Sigma-Aldrich

Pinoresinol diglucoside

≥98% (HPLC)

Synonym(s):

(+)-Pinoresinol 4,4′-O-bisglucopyranoside, Pinoresinol di-β-D-glucoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H42O16
CAS Number:
Molecular Weight:
682.67
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98% (HPLC)

form

powder

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O[C@H]1[C@H](O)[C@@H](CO)O[C@@H](OC2=CC=C([C@H]3OC[C@@]4([H])[C@]3([H])CO[C@@H]4C5=CC=C(O[C@@H]6O[C@H](CO)[C@@H](O)[C@H](O)[C@H]6O)C(OC)=C5)C=C2OC)[C@@H]1O

InChI

1S/C32H42O16/c1-41-19-7-13(3-5-17(19)45-31-27(39)25(37)23(35)21(9-33)47-31)29-15-11-44-30(16(15)12-43-29)14-4-6-18(20(8-14)42-2)46-32-28(40)26(38)24(36)22(10-34)48-32/h3-8,15-16,21-40H,9-12H2,1-2H3/t15-,16-,21+,22+,23+,24+,25-,26-,27+,28+,29+,30+,31+,32+/m0/s1

InChI key

ZJSJQWDXAYNLNS-FUPWJLLWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pinoresinol diglucoside (PDG) is an important lignan found in the bark of the traditional Chinese medicinal plant Eucommia ulmoides, also known as Tu-Chung. It is also associated with other plants such as Forsythia suspensa and Styrax spp. In plants, PDG is produced through the phenylpropanoid pathway.
Pinoresinol diglucoside is an important lignan found in the bark of the Eucommia ulmoides, which is known in traditional Chinese medicine as du zhong. In plants, pinoresinol diglucoside is produced through the phenylpropanoid pathway.

Biochem/physiol Actions

Pinoresinol diglucoside has antihypertensive properties. It serves as a α-glucosidase inhibitor. α-Glucosidase catalyzes the conversion of carbohydrates to glucose in the small intestine. Hence, pinoresinol diglucoside might be useful as an antidiabetic agent.
Pinoresinol diglucoside is a potent antihypertensive, anti-inflammatory and antioxidant agent. It also exerts tumor-suppressive and anti-osteoporotic properties. PDG serves as an α-glucosidase inhibitor that catalyzes the conversion of carbohydrates to glucose in the small intestine. Hence, it may serve as an antidiabetic agent in the management of diabetes. It displays neuroprotective properties by alleviating ischemic stroke-related brain injury in mice model.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhenhong Gao et al.
Journal of microbiology and biotechnology, 27(8), 1428-1440 (2017-06-18)
Phomopsis sp. XP-8 (an endophytic fungus) was previously found to produce pinoresinol diglucoside (PDG), a major antihypertensive compound of Tu-Chung (the bark of Eucommia ulmoides Oliv.), which is widely used in Chinese traditional medicines. In the present study, two bioconversion
M Bulat et al.
The Journal of physiology, 275, 191-197 (1978-02-01)
1. After inhibition of 5-hydroxyindoleacetic acid (5-HIAA) transport by repeated administration of probenecid in cats, this acid accumulates linearly in the lumbosacral cord, lumbar cerebrospinal fluid (c.s.f.) and cisternal fluid during 5 hr of experiment. 2. A simple mathematical analysis
M Harada et al.
Hinyokika kiyo. Acta urologica Japonica, 38(2), 207-211 (1992-02-01)
A rare case of ureteral endometriosis is reported. A 46-year-old nulliparous woman was referred to our hospital because of right hydronephrosis which was detected by ultrasonography; Drip infusion pyelography showed right hydroureteronephrosis with the distal obstruction. MRI demonstrated right ovarian
Bioconversion of pinoresinol diglucoside and pinoresinol from substrates in the phenylpropanoid pathway by resting cells of Phomopsis sp. XP-8
Zhang Y, et al.
PLoS ONE, 10(9), e0137066-e0137066 (2015)
J Lukl et al.
Vnitrni lekarstvi, 38(1), 1-5 (1992-01-01)
In the course of nine years the authors investigated in 3570 patients hospitalized on account of acute coronary syndrome associations between unstable angina pectoris (UAP) and a acute myocardial infarction (NMI). A total of 1732 patients were admitted with UAP

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service