Skip to Content
Merck
All Photos(1)

Documents

SMB00400

Sigma-Aldrich

Ligustilide

≥96% (HPLC)

Synonym(s):

3-butylidene-4,5-dihydro-1(3H)-isobenzofuranone, 3-butylidene-4,5-dihydrophthalide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14O2
CAS Number:
Molecular Weight:
190.24
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥96% (HPLC)

form

liquid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O=C(O/C1=C\CCC)C2=C1CCC=C2

InChI

1S/C12H14O2/c1-2-3-8-11-9-6-4-5-7-10(9)12(13)14-11/h5,7-8H,2-4,6H2,1H3/b11-8-

InChI key

IQVQXVFMNOFTMU-FLIBITNWSA-N

General description

Ligustilide is a phthalide derivative and is one of the main lipophilic compounds of Danggui essential oil and Rhizoma Chuanxiong, a Chinese medicinal herb.

Application

Ligustilide has been used as a treatment to test its anti-apoptotic effects on rat chondrocytes and sodium nitroprusside (SNP)-stimulated chondrocytes in both in vitro and in vivo studies. It has also been used as a treatment to study anti-atherosclerosis effects on high-fat diet-fed mice and cytoprotective effects on tert-butyl hydroperoxide (t-BHP)-evoked EA.hy926 cells.

Biochem/physiol Actions

Ligustilide can inhibit the proliferation of and cell cycle progression of vascular smooth muscle cells and inhibits vasoconstriction. It increases vasodilation, anti-thrombosis, and serotonergic activity and decreases platelet aggregation. It has been shown to have broad applications in the study and treatment of cardiovascular diseases and ischemic brain injury. Ligustilide has the ability to reduce neurogenic and inflammatory pain. Thus, it can be used as a potential therapeutic agent to treat several pain syndromes including primary dysmenorrhoea.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ligustilide ameliorates neuroinflammation and brain injury in focal cerebral ischemia/reperfusion rats: involvement of inhibition of TLR4/peroxiredoxin 6 signaling.
Kuang X, et al.
Free Radical Biology & Medicine, 71, 165-175 (2014)
Pharmacokinetics and Metabolism of Ligustilide, a Major Bioactive Component in Rhizoma Chuanxiong, in the Rat
Yan R, et al.
Drug Metabolism and Disposition (2007)
Ligustilide attenuates pain behavior induced by acetic acid or formalin
Du J, et al.
Journal of Ethnopharmacology, 112(1), 211-214 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service