Skip to Content
Merck
All Photos(3)

Documents

SMB00313

Sigma-Aldrich

Gastrodin

≥98% (HPLC)

Synonym(s):

(2R,3S,4S,5R,6S)-2-Hydroxymethyl-6-(4-hydroxymethyl-phenoxy)-tetrahydro-pyran-3,4,5-triol, 4-β-D-glucopyranosyloxybenzyl alcohol, 4-Hydroxybenzyl alcohol 4-O-β-D-glucoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H18O7
CAS Number:
Molecular Weight:
286.28
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

biological source

plant (Gastrodia elata Blume)

Quality Level

Assay

≥98% (HPLC)

form

powder

application(s)

detection
metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

O[C@@H]1[C@@H](CO)O[C@@H](OC2=CC=C(CO)C=C2)[C@H](O)[C@H]1O

InChI

1S/C13H18O7/c14-5-7-1-3-8(4-2-7)19-13-12(18)11(17)10(16)9(6-15)20-13/h1-4,9-18H,5-6H2/t9-,10-,11+,12-,13-/m1/s1

InChI key

PUQSUZTXKPLAPR-UJPOAAIJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Gastrodin, a phenolic glucoside, is the main active ingredient of the traditional Chinese medicinal compound Tianma. Gastrodin is an active component of Gastrodia elata Blume, a herb belonging to the Orchidaceae family.

Application

Gastrodin has been used as a control to estimate the gastrodin content in the leaves, roots and stems of Anoectochilus formosanus Hayata.

Biochem/physiol Actions

Gastrodin can penetrate through the blood-brain barrier and has been suggested to have neuroprotective effects against hypoxia. It is also known to reduce the immunoreactivity of gamma-aminobutyric acid shunt enzymes in hippocampal tissues. Traditionally, gastrodin is also known to be effective in the treatment of headache, vertigo, dizziness and convulsive disorders. Along with anticonvulsant property, it also possesses antiepileptic, anxiolytic, anti-inflammatory and antiobesity action. Gastrodin is also helps in learning and memory stimulation.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enhanced Agronomic Traits and Medicinal Constituents of Autotetraploids in Anoectochilus formosanus Hayata, a Top-Grade Medicinal Orchid
Chung HH, et al.
Molecules (Basel), 22(11), 1907-1907 (2017)
Gastrodin protects apoptotic dopaminergic neurons in a toxin-induced Parkinson?s disease model
Kumar H, et al.
Evidence-Based Complementary and Alternative Medicine : ECAM, 2013 (2013)
Yu Xing et al.
The international journal of biochemistry & cell biology, 109, 8-16 (2019-01-27)
The in vivo protective role of gastrodin (GSTD) in myocardial infarction (MI) has been reported. However, the underlying mechanism remains unclear. Herein, we aimed to explore the effects of GSTD on hypoxia-injured H9c2 cells as well as the downstream microRNAs
Tianyuan Ye et al.
International journal of molecular sciences, 19(12) (2018-12-14)
Patients with diabetes mellitus (DM) suffer more risks from diabetic encephalopathy such as cognitive dysfunction and depressive-like behaviors. Numerous studies show that ER (endoplasmic reticulum) stress and inflammation play important roles in the development of diabetic encephalopathy. Gastrodin (Gas), one
Man Jiao et al.
Chemosphere, 226, 678-686 (2019-04-09)
The ubiquitous environmental pollutant 2,3,7,8-tetrachlorodibenzo-p-dioxin (TCDD) has been shown to trigger neurotoxicity. In this study, we investigated the protective effects of gastrodin on TCDD-induced neurotoxicity and the underlying molecular mechanisms. The results show that gastrodin decreased cell viability, tumor necrosis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service