Skip to Content
Merck
All Photos(1)

Key Documents

SCP0257

Sigma-Aldrich

Tryptase Substrate

≥95% (HPLC), lyophilized

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H42N8O6
Molecular Weight:
634.73
UNSPSC Code:
12352204
NACRES:
NA.32

product name

Tryptase Substrate,

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥65%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Ac-Orn-Phe-Arg-AMC

Application

Tryptase is a serine proteinase often used as a marker for mast cell activation. Tryptase is detected using peptides such as acetyl-Orn-Phe-Arg-AMC, a fluorogenic substrate.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service