Skip to Content
Merck
All Photos(1)

Key Documents

SAE0160

Sigma-Aldrich

Albumin human

Exbumin, Recombinant, lyophilized powder, Animal Component Free, suitable for cell culture, low endotoxin

Synonym(s):

ExbuminInVitria, rHSA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.25

biological source

human

Quality Level

form

powder

mol wt

monomer ~67 kDa

concentration

>96%

technique(s)

cell culture | mammalian: suitable

impurities

Endotoxin, tested

shipped in

wet ice

storage temp.

−20°C

General description

Exbumin is high-purity recombinant human serum albumin with high lysine reactivity. It contains low levels of albumin aggregates and lipids and high concentration of free-thiol/Cys-34 mercapto-albumin. This USA-origin rHSA is animal-free.

Application

ExbuminInvitria is recombinant human serum that acts as carrier protein for fatty acids and growth factors. It is added as an ingredient to classical and custom media formulations. This recombinant variant is recommended for use in absorption, distribution, metabolism, and excretion (ADME) pharmacological research; cell culture; drug delivery research; and cryopreservation of cells. ExbuminInvitria has been observed to improve the performance of multiple cell types.

Biochem/physiol Actions

Serum albumin functions as a carrier protein for steroids, fatty acids, and thyroid hormones. They are also vital in regulating the colloidal osmotic pressures of blood. Serum albumins protect the cells against physical or toxins-induced stress.

Features and Benefits

Exbumin is animal component free that is manufactured using good manufacturing practices at an ISO9001:2015 certified facility. Exbumin is also used to maintain tissue viability during cryo-preservation or storage.
Advantages


  • Eliminates animal-derived additives
  • Safe and regulatory friendly
  • Provides well-defined and consistent stem cell medium
  • Raw materials, manufacturing equipment, and packaging is certified blood free and animal free

Packaging

5 g in poly bottle

Preparation Note

To prepare stock solutions, gently dissolve Exbumin in DPBS, PBS or basal media at a concentration of 50-100mg/mL. If needed, allow Exbumin to completely dissolve overnight at 4°C in a dark environment. Filter sterilization with a 0.8μm filter followed by a 0.2μm filter is recommended. Liquid solutions can be stored for 4 weeks at 4°C in the dark.

Other Notes

Exbumin is a trademark of InVitria

Legal Information

Exbumin is a trademark of Ventria Bioscience Inc.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service