Skip to Content
Merck
All Photos(3)

Key Documents

SAB5500168

Sigma-Aldrich

Anti-PTEN antibody, Rabbit monoclonal

recombinant, expressed in proprietary host, clone SP170, affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in proprietary host

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

SP170, monoclonal

species reactivity

human (tested)

species reactivity (predicted by homology)

dog, mouse

technique(s)

flow cytometry: 1:400
immunoblotting: 1:100
immunohistochemistry: 1:100

isotype

IgG

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... PTEN(5728)

Related Categories

General description

Phosphatidylinositol-3, 4, 5-trisphosphate 3-phosphatase and dual-specificity protein phosphatase (PTEN) is a tumor suppressor and a member in the PI3K/PTEN/AKT pathway. PTEN is a phosphatase with dual specificity to slow cell growth and antagonize the growth promoting activities from HER protein kinases. The defects of PTEN, mostly around the C-terminus of PTEN, have been implicated in many human cancers and diseases. Therefore, antibody to the N terminus will not be able to detect most of the PTEN protein loss.

Immunogen

Synthetic peptide close to the N-terminus of human PTEN protein.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

0.1 ml rabbit monoclonal antibody purified by protein A/G in PBS/1% BSA buffer pH 7.6 with less than 0.1% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service