Skip to Content
Merck
All Photos(3)

Key Documents

SAB4503896

Sigma-Aldrich

Anti-phospho-AS160 (pThr642) antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 146 kDa

species reactivity

human, mouse, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pThr642)

Gene Information

human ... TBC1D4(9882)

Immunogen

The antiserum was produced against synthesized peptide derived from human AS160 around the phosphorylation site of Thr642.

Immunogen Range: 611-660

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hyo-Bum Kwak et al.
International journal of environmental research and public health, 16(7) (2019-04-03)
In rodent skeletal muscle, acyl-coenzyme A (CoA) synthetase 5 (ACSL-5) is suggested to localize to the mitochondria but its precise function in human skeletal muscle is unknown. The purpose of these studies was to define the role of ACSL-5 in
Xuan He et al.
Chinese medical journal, 132(14), 1713-1722 (2019-07-04)
Macrophage accumulation in the vascular wall is a hallmark of atherosclerosis. Studies showed that shifting of oxidized lipids-induced inflammatory macrophages towards an anti-inflammatory phenotype by promoting oxidative metabolism attenuated atherosclerosis progression. Therefore, this study aimed to investigate whether metformin, which
Shuang Peng et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 58, 152745-152745 (2019-04-22)
Rare ginsenosides are found in ginseng and notoginseng, two medicinal plants widely used in China for treatment of cardiovascular diseases and type 2 diabetes. However, their pharmacological studies regarding myocardial fuel metabolism and insulin signaling are not clear. To explore
Filippe Falcão-Tebas et al.
The Journal of physiology, 597(1), 121-136 (2018-11-09)
A paternal high-fat diet/obesity before mating can negatively influence the metabolism of offspring. Exercise only early in life has a remarkable effect with respect to reprogramming adult rat offspring exposed to detrimental insults before conception. Exercise only early in life
Edward B Arias et al.
PloS one, 14(4), e0216236-e0216236 (2019-04-30)
Akt substrate of 160 kDa (also called AS160 or TBC1D4) is a Rab GTPase activating protein and key regulator of insulin-stimulated glucose uptake which is expressed by multiple tissues, including skeletal muscle, white adipose tissue (WAT) and the heart. This

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service