Skip to Content
Merck
All Photos(4)

Key Documents

SAB4503882

Sigma-Aldrich

Anti-phospho-IKK-α (pSer176) /IKK-β (pSer177) antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 84 kDa

species reactivity

rat, human, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer176/pSer177)

Gene Information

General description

CHUK (conserved helix-loop-helix ubiquitous kinase) is located on human chromosome 10q24. IKK1/IKK-α has three ordered protein domains, namely, a kinase domain (KD), a ubiquitin-like domain (ULD) and a scaffold dimerization domain (SDD). IKKB (inhibitor of nuclear factor κ B kinase subunit β) is mapped to human chromosome 8p11. IKBKB codes for IκB kinase 2 (IKK2/IKKβ). IKK1 and IKK2 has a canonical MAP kinase kinase (MAPKK) activation loop motif.

Immunogen

The antiserum was produced against synthesized peptide derived from human IKK-alpha around the phosphorylation site of Ser177.

Immunogen Range: 151-200

Biochem/physiol Actions

IKK1/IKKα (I-κ-B kinase 1) modulates nuclear factor κ-B (NF-κB) signaling, that is required for lymphoid organogenesis and adaptive immunity. IκB kinase α (IKKα) plays a major role in human skin cancer. This gene also controls terminal differentiation and multiplication of keratinocytes. IKKβ acts as the master regulator of cell survival and the inflammatory response.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structural Basis for the Activation of IKK1/α.
Polley S, et al.
Cell Reports, 17(8), 1907-1914 (2016)
Chapter 252 - NFκB: A Key Integrator of Cell Signaling
Handbook of Cell Signaling, 2069-2076 (2009)
I?B kinase α functions as a tumor suppressor in epithelial-derived tumors through an NF-κB-independent pathway.
Xie Y, et al.
Oncology Reports, 34(5), 2225-2232 (2015)
IKKβ promotes metabolic adaptation to glutamine deprivation via phosphorylation and inhibition of PFKFB3.
Reid M A, et al.
Genes & Development, 30(16), 1837?1851-1837?1851 (2016)
A suite of novel human spermatozoal RNAs.
Ostermeier G C, et al.
Journal of Andrology, 26(1), 70-74 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service