Skip to Content
Merck
All Photos(2)

Key Documents

SAB4503363

Sigma-Aldrich

Anti-ACSS1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Acetate--CoA ligase 2, Acetyl-CoA synthetase 2, Acetyl-coenzyme A synthetase 2-like, Acyl-CoA synthetase short-chain family member 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
ELISA
IHC
WB
clone:
polyclonal
species reactivity:
human, mouse
citations:
3
technique(s):
ELISA: 1:40000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 74 kDa

species reactivity

human, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:40000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ACSS1(84532)

General description

Anti-ACSS1 Antibody detects endogenous levels of total ACSS1 protein.
Acetyl coenzyme A (CoA) synthetase 1 (ACSS1) is an enzyme which is also termed as Acecs2. It is present in the mitochondria. It is located on human chromosome 20p11.21.

Immunogen

The antiserum was produced against synthesized peptide derived from human ACSS1.

Immunogen Range: 611-660

Biochem/physiol Actions

Acetyl coenzyme A (CoA) synthetase 1 (ACSS1) participates in the oxidation of acetate. It converts acetate to acetyl-CoA in an ATP-dependent reaction. ACSS1 promotes aggressive tumor growth.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jagged1 DNA Copy Number Variation Is Associated with Poor Outcome in Liver Cancer.
Kawaguchi K, et al.
The American Journal of Pathology, 186(8), 2055-2067 (2016)
Glucose-independent Acetate Metabolism Promotes Melanoma Cell Survival and Tumor Growth.
Lakhter AJ, et al.
The Journal of Biological Chemistry, 291(42), 21869-21879 (2016)
Acetyl-CoA synthetase 2, a mitochondrial matrix enzyme involved in the oxidation of acetate.
Fujino T, et al.
The Journal of Biological Chemistry, 276(14), 11420-11426 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service