Skip to Content
Merck
All Photos(1)

Documents

SAB4200742

Sigma-Aldrich

Anti-c-Myc-Peroxidase antibody, Mouse monoclonal

clone 9E10, purified from hybridoma cell culture

Synonym(s):

Anti-MRTL, Anti-Myc, Anti-bHLHe39, Anti-v-myc avian myelocytomatosis viral oncogene homolog

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.56

biological source

mouse

Quality Level

conjugate

peroxidase conjugate

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

9E10, monoclonal

form

lyophilized powder

species reactivity

human

packaging

vial of 100 μL

concentration

~2 mg/mL

technique(s)

immunoblotting: 1:250-1:500 using lysate of HEK-293T cells over expressing c-Myc fusion protein
immunohistochemistry: suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MYC(4609)

General description

Monoclonal Anti-c-Myc (mouse IgG1 isotype) is derived from the 9E10 hybridoma, produced by the fusion of mouse myeloma cells and splenocytes from a BALB/c mouse immunized with synthetic peptide of the human p62c-myc protein, conjugated to KLH. c-Myc is a proto-oncogene localized on long arm of human chromosome 8. The encoded protein contains an N-terminal transactivation domain. The human c-Myc proto-oncogene is the cellular homolog of the avian v-Myc gene found in several leukemogenic retroviruses.

Specificity

Monoclonal Anti-Calponin specifically recognizes smooth muscle Calponin.

Immunogen

Synthetic peptide of the human p62c-myc protein.

Application

Anti-c-Myc-Peroxidase antibody, Mouse monoclonal may be used in immunoblotting and immunohistology.

Biochem/physiol Actions

c-Myc is a transcription factor involved in regulation of cell growth, proliferation, differentiation and apoptosis. It is implicated in DNA replication, RNA splicing or transcription. Increased expression of the gene is associated with the development of various types of human cancers.

Physical form

Supplied as lyophilized powder. After reconstitution with 0.1 mL of distilled water to a final antibody concentration of ∼ 2 mg/mL, the solution contains 1% BSA, 2.5% trehalose, 0.05% MIT in 0.01 M sodium phosphate buffered saline

Storage and Stability

Store the lyophilized product at 2–8 °C. For extended storage after reconstitution, keep at –20 °C in working aliquots. Avoid repeated freeze-thaw cycles. For continuous use after reconstitution, keep at 2–8 °C for up to 1 month. Solutions at working dilution should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog, our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The c-Myc target gene network
Dang CV, et al.
Seminars in Cancer Biology, 16(4), 253-264 (2006)
Prognostic Value of c-Myc Immunohistochemical Expression in Muscle Invasive Urothelial Carcinoma of the Urinary Bladder: A Retrospective Study
Elwy AE, et al.
Asian Pacific Journal of Cancer Prevention, 20(12), 3735-3735 (2019)
Hainan Liu et al.
Journal of virology, 96(12), e0041222-e0041222 (2022-06-03)
SARS-CoV-2 is the causative agent of the ongoing pandemic of coronavirus disease 2019 (COVID-19) and poses a significant threat to global health. N protein (NP), which is a major pathogenic protein among betacoronaviruses, binds to the viral RNA genome to
Tong Zheng et al.
Journal of virology, 98(4), e0156523-e0156523 (2024-03-06)
The COVID-19 pandemic caused by severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) has posed a worldwide threat in the past 3 years. Although it has been widely and intensively investigated, the mechanism underlying the coronavirus-host interaction requires further elucidation, which
Lin Zhu et al.
eLife, 12 (2024-01-29)
Viral inclusion bodies (IBs) commonly form during the replication of Ebola virus (EBOV) in infected cells, but their role in viral immune evasion has rarely been explored. Here, we found that interferon regulatory factor 3 (IRF3), but not TANK-binding kinase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service