Skip to Content
Merck
All Photos(1)

Key Documents

SAB2500236

Sigma-Aldrich

Anti-Choline Acetyltransferase Antibody

goat polyclonal

Synonym(s):

Anti-Acetyl CoA:choline O-acetyltransferase, Anti-CHAT, Anti-CMS1A, Anti-CMS1A2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

Product Name

Anti-Choline acetyltransferase antibody produced in goat, affinity isolated antibody, buffered aqueous solution

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

canine, chimpanzee, human, pig

technique(s)

indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CHAT(1103)

General description

Choline acetyltransferase (CHAT) has a molecular weight of about 68kDa. It is synthesized in the parakaryon and is transported to the axon terminals. CHAT is part of the choline/carnitine acyltransferase family.

Immunogen

Peptide with sequence CKEKATRPSQGHQP from the C Terminus of the protein sequence according to NP_065574; NP_066264; NP_066265; NP_066266.

Biochem/physiol Actions

Choline acetyltransferase (CHAT) is involved in the synthesis of acetylcholine in neurons. It catalyzes the transfer of an acetyl group between acetyl-coenzyme A and choline. Mutations in the gene encoding this protein have been associated with congenital neuromuscular disorders.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Blood pressure regulation by CD4+ lymphocytes expressing choline acetyltransferase.
Olofsson PS, e.al
Nature Biotechnology (2016)
Choline acetyltransferase structure reveals distribution of mutations that cause motor disorders
The Embo Journal (2004)
Francesco Antoniani et al.
Cell death discovery, 9(1), 248-248 (2023-07-16)
Amyotrophic Lateral Sclerosis (ALS) and Frontotemporal Dementia (FTD) are two neurodegenerative disorders that share genetic causes and pathogenic mechanisms. The critical genetic players of ALS and FTD are the TARDBP, FUS and C9orf72 genes, whose protein products, TDP-43, FUS and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service