Skip to Content
Merck
All Photos(4)

Key Documents

SAB2104153

Sigma-Aldrich

Anti-ZFR antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-FLJ41312

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

117 kDa

species reactivity

bovine, rat, mouse, horse, human, rabbit, dog

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZFR(51663)

General description

Zinc finger RNA binding protein (ZFR) is a highly conserved protein and is homologous to mouse ZFR. It is essential in the early developmental stages. ZFR has three N-terminal C2H2 zinc finger motifs and a C-terminal DZF (domain associated with zinc fingers) domain. ZFR is highly expressed in brain, whereas skeletal muscles is devoid of ZFR expression. In human chromosome, the gene ZFR is localized on 5p13.3.

Immunogen

Synthetic peptide directed towards the middle region of human ZFR

Application

Anti-ZFR antibody produced in rabbit has been used in western blot analysis.

Biochem/physiol Actions

Zinc finger RNA binding protein (ZFR) binds to RNA and DNA and promotes DNA repair and chromosome organisation. ZFR regulates alternative pre-mRNA splicing and suppress interferon 1 expression in tissues. ZFR is a key factor in regulating transcription in macrophages. ZFR is critical for Staufen 2 isoform specific nucleocytoplasmic shuttling in neurons. ZFR is a potent marker and therapeutic target in cancer. ZFR is highly expressed in pancreatic cancer and knockdown of which suppressed the tumor cell growth and proliferation. ZFR is overexpressed in non-small-cell lung carcinoma (NSCLC) and causes cell growth and metastasis.

Sequence

Synthetic peptide located within the following region: RQIHKVLGMDPLPQMSQRFNIHNNRKRRRDSDGVDGFEAEGKKDKKDYDN

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Knockdown of ZFR suppresses cell proliferation and invasion of human pancreatic cancer
Zhao X, et al.
Biological Research, 49(1), 26-26 (2016)
Identification of a locus for autosomal dominant high myopia on chromosome 5p13. 3-p15. 1 in a Chinese family
Ma JH, et al.
Molecular Vision, 16(1), 2043-2043 (2010)
ZFR coordinates crosstalk between RNA decay and transcription in innate immunity
Haque N, et al.
Nature Communications, 9(1), 1145-1145 (2018)
DHX9 suppresses RNA processing defects originating from the Alu invasion of the human genome
Aktas T, et al.
Nature, 544(7648), 115-115 (2017)
Xiaolan Zhao et al.
Biological research, 49(1), 26-26 (2016-05-15)
Zinc finger RNA binding protein (ZFR) is involved in the regulation of growth and cancer development. However, little is known about ZFR function in pancreatic cancer. Herein, to investigate whether ZFR is involved in tumor growth, Oncomine microarray data was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service