SAB1403022
Monoclonal Anti-PEX16 antibody produced in mouse
clone 3E10, ascites fluid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
Recommended Products
biological source
mouse
conjugate
unconjugated
antibody form
ascites fluid
antibody product type
primary antibodies
clone
3E10, monoclonal
mol wt
antigen ~64.17 kDa
species reactivity
human
technique(s)
indirect ELISA: suitable
isotype
IgMκ
NCBI accession no.
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... PEX16(9409)
General description
The protein encoded by this gene is an integral peroxisomal membrane protein. An inactivating nonsense mutation localized to this gene was observed in a patient with Zellweger syndrome of the complementation group CGD/CG9. Expression of this gene product morphologically and biochemically restores the formation of new peroxisomes, suggesting a role in peroxisome organization and biogenesis. Alternative splicing has been observed for this gene and two variants have been described. (provided by RefSeq)
Immunogen
PEX16 (AAH00467, 1 a.a. ~ 346 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Sequence
MEKLRLLGLRYQEYVTRHPAATAQLETAVRGFSYLLAGRFADSHELSELVYSASNLLVLLNDGILRKELRKKLPVSLSQQKLLTWLSVLECVEVFMEMGAAKVWGELGRWLVIALIQLAKAVLRILLLLWFKAGLQTSPPIAPLDRETQAQPPDGDHSPGNHEQSYVGKRSNRVVRTLQNTPSLHSRHWGAPQQREGRQQQHHEELSATPTPLGLQETIAEFLYIARPLLHLLSLGLWGQRSWKPWLLAGVVDVTSLSLLSDRKGLTRRERRELRRRTILLLYYLLRSPFYDRFSEARILFLLQLLADHVPGVGLVTTSQRAASPCLPARPHTQPWSPPAFLPGHP
Sequence
MEKLRLLGLRYQEYVTRHPAATAQLETAVRGFSYLLAGRFADSHELSELVYSASNLLVLLNDGILRKELRKKLPVSLSQQKLLTWLSVLECVEVFMEMGAAKVWGELGRWLVIALIQLAKAVLRILLLLWFKAGLQTSPPIAPLDRETQAQPPDGDHSPGNHEQSYVGKRSNRVVRTLQNTPSLHSRHWGAPQQREGRQQQHHEELSATPTPLGLQETIAEFLYIARPLLHLLSLGLWGQRSWKPWLLAGVVDVTSLSLLSDRKGLTRRERRELRRRTILLLYYLLRSPFYDRFSEARILFLLQLLADHVPGVGLVTTSQRAASPCLPARPHTQPWSPPAFLPGHP
Physical form
Clear solution
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service