S2257
Sera human
lyophilized powder (from clotted whole blood)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
human
form
lyophilized powder (from clotted whole blood)
origin
USA origin
storage temp.
2-8°C
Application
Not tested for use in cell culture
Physical form
0.2μ filtered prior to lyophilization from the indicated amount
Disclaimer
RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
The Analyst, 141(20), 5730-5737 (2016-07-08)
An electrochemical immunosensor for the determination of the multifunctional Transforming Growth Factor β1 (TGF-β1) cytokine has been prepared using multi-walled carbon nanotube (MWCNT)-modified screen-printed carbon electrodes. MWCNTs were functionalized by means of copper(i) catalyzed azide-alkyne cycloaddition ("click" chemistry) as an
Analytica chimica acta, 1061, 84-91 (2019-03-31)
We report herein the design of a novel biosensing strategy for the detection of carcinoembryonic antigen (CEA), based on the use of Janus-type nanoparticles having Au and silica opposite faces as integrated electrochemical biorecognition-signaling system. The Janus nanoparticles were properly
Analytica chimica acta, 959, 66-73 (2017-02-06)
Dual screen-printed carbon electrodes modified with 4-carboxyphenyl-functionalized double-walled carbon nanotubes (HOOC-Phe-DWCNTs/SPCEs) have been used as scaffolds for the preparation of electrochemical immunosensors for the simultaneous determination of the cytokines Interleukin-1β (IL-1β) and factor necrosis tumor α (TNF-α). IL-1β. Capture antibodies
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service