Key Documents
RFA452411K-EP
Rotor for Eppendorf® 5430/5430R Centrifuges
FA-45-24-11-Kit (rotor with higher edge), aerosol-tight, to hold, 24 x spin columns, lid
About This Item
UNSPSC Code:
41103900
feature
lid
to hold 24 x spin columns
aerosol-tight
fixed angle rotor
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 022654128 (NA)
Eppendorf® 5427752005 (EU)
parameter
19,090 × g max. RCF
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5430/5430R
General description
Rotors to fot Eppendorf centrifuge 5430 and 5430 R.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)