RAB0227
Rat IFN-γ ELISA Kit
for serum, plasma and cell culture supernatant
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41116158
NACRES:
NA.32
Recommended Products
species reactivity
rat
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
capture ELISA: suitable
input
sample type cell culture supernatant(s)
sample type serum
sample type plasma
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 100 pg/mL
standard curve range: 122.9-30000 pg/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
rat ... Ifng(25712)
General description
The Rat IFN-γ ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of Rat IFN-γ in serum, plasma, cell culture supernatants and urine.
Immunogen
Recombinant Rat IFN-γ
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Other Notes
A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.
Please type the word sample in the text box provided for lot number.
Kit Components Also Available Separately
Product No.
Description
SDS
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
K Kim et al.
Tuberculosis (Edinburgh, Scotland), 94(5), 494-501 (2014-08-07)
Previous studies suggest that control of Mycobacterium tuberculosis infection is compromised by the activity of regulatory T cells, including those that express CD39, an ectonucleotidase with immunosuppressive properties. Here, we examine the role of CD39 on CD4+ T cells reacting
Kaveh Abdi et al.
Journal of immunology (Baltimore, Md. : 1950), 192(12), 6028-6036 (2014-05-14)
IL-12p40 partners with the p35 and p19 polypeptides to generate the heterodimeric cytokines IL-12 and IL-23, respectively. These cytokines play critical and distinct roles in host defense. The assembly of these heterodimers is thought to take place within the cell
Hamid Reza Jafarinaveh et al.
Iranian journal of allergy, asthma, and immunology, 13(2), 125-130 (2013-12-18)
Increased levels of proinflammatory cytokines have been recorded after the onset of transient or permanent brain ischemia and are usually associated with exacerbation of ischemic injury. Embolic stroke model is more relevant to the pathophysiological situation in such patients, because
Xiaowei Wu et al.
Chemistry, an Asian journal, 9(8), 2260-2271 (2014-04-05)
We have previously described the discovery of N-alkylated iminosugars that showed immunosuppressive activity both in vitro and in vivo. Herein, we report the synthesis and biological evaluation of N-arylated lactam-type iminosugar derivatives. The synthesis started from simple monosaccharides and featured a Buchwald-Hartwig
Chris P Verschoor et al.
Infection and immunity, 82(11), 4824-4833 (2014-08-27)
Pneumonia caused by Streptococcus pneumoniae is a major cause of death and an economic burden worldwide. S. pneumoniae is an intermittent colonizer of the human upper respiratory tract, and the ability to control asymptomatic colonization determines the likelihood of developing
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service