Skip to Content
Merck
All Photos(1)

Key Documents

PZ0215

Sigma-Aldrich

PF-04701475

≥98% (HPLC)

Synonym(s):

N-[[(5S)-3-[3-Fluoro-4-(1-pyrrolidinyl)phenyl]-4,5-dihydro-5-isoxazolyl]methyl]-2-propanesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H24FN3O3S
CAS Number:
Molecular Weight:
369.45
UNSPSC Code:
51111800
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear

storage temp.

room temp

Biochem/physiol Actions

PF-04701475 is a potent and selective α-Amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid receptor (AMPAR) positive allosteric potentiator.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service