Skip to Content
Merck
All Photos(2)

Key Documents

PRS2301

Sigma-Aldrich

Anti-AIF antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Connector enhancer of kinase suppressor of Ras 1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
ELISA (i)
ICC
WB
clone:
polyclonal
species reactivity:
mouse, human, rat
citations:
2
technique(s):
immunocytochemistry: suitable
indirect ELISA: suitable
western blot: suitable

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 67 kDa

species reactivity

mouse, human, rat

technique(s)

immunocytochemistry: suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CNKSR1(10256)

General description

The previously assigned protein identifier O95381 has been merged into Q969H4. Full details can be found on the UniProt database.

Immunogen

a peptide corresponding to amino acids 593-606 of human AIF. This sequence is identical to mouse and rat AIF.

Linkage

The action of this antibody can be blocked using blocking peptide SBP2301.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaowan Wang et al.
International journal of molecular sciences, 15(11), 20449-20468 (2014-11-12)
NAD+ is an essential co-enzyme for cellular energy metabolism and is also involved as a substrate for many cellular enzymatic reactions. It has been shown that NAD+ has a beneficial effect on neuronal survival and brain injury in in vitro
Xiaowan Wang et al.
Journal of neurochemistry, 151(6), 732-748 (2019-09-26)
NAD+ is a cofactor required for glycolysis, tricarboxylic acid cycle, and complex I enzymatic reaction. In mammalian cells, NAD+ is predominantly synthesized through the salvage pathway, where nicotinamide phosphoribosyltransferase (NAMPT) is the rate-limiting enzyme. Previously, we demonstrated that NAMPT exerts

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service