MAK222
Beta-Lactamase Inhibitor Screening Kit
Sufficient for 100 Colorimetric tests
Synonym(s):
β-Lactamase Inhibitor Screening Kit
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
12161503
NACRES:
NA.84
Recommended Products
detection method
colorimetric
storage temp.
−20°C
General description
β-Lactamase (β-lactamase, βL, EC 3.5.2.6) is an enzyme first identified in Escherichia coli and has been described as penicillinase. A number of βLs have since been identified from various bacteria. β-Lactamases specifically hydrolyze β-lactam rings present in antibiotics such as penicillin, cephalosporins, monobactam, and carbapenem, and confer resistance against these antibiotics.
Application
Beta-Lactamase Inhibitor Screening Kit has been used in enzymatic assays.
Features and Benefits
Compatible with high-throughput handling systems.
Suitability
Suitable for the screening of inhibitors of β-Lactamase
Principle
The β-Lactamase Inhibitor Screening Kit is a rapid, simple and sensitive assay that is suitable for high throughput screening of β-Lactamase inhibitors. βL activity is measured by hydrolyzing a chromogenic cephalosporin called nitrocefin, producing a colorimetric product (A490), proportional to the enzymatic activity present.
Kit Components Only
Product No.
Description
- β-Lactamase Assay Buffer
- Nitrocefin, in DMSO
- β-Lactamase
- Inhibitor Control, Clavulanic acid
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Resp. Sens. 1 - Skin Sens. 1
Storage Class Code
10 - Combustible liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Evaluation of 1, 4, 7-Triazacyclononane (TACN) as a potential Metallo-B-Lactamase inhibitor in Enterobacteriaceae: Restoring the Activity of B-lactams
Somboro A M, et al.
bioRxiv, 366146-366146 (2018)
Anou M Somboro et al.
Applied and environmental microbiology, 85(3) (2018-11-28)
Metallo-β-lactamase (MBL)-producing Enterobacteriaceae are of grave clinical concern, particularly as there are no metallo-β-lactamase inhibitors approved for clinical use. The discovery and development of MBL inhibitors to restore the efficacy of available β-lactams are thus imperative. We investigated a zinc-chelating
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service