M0938
7-Methoxycoumarin-4-Acetyl-Gly-Lys-Pro-Ile-Leu-Phe-Phe-Arg-Leu-Lys(DNP)-D-Arg-amide
>97% (HPLC), solid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C85H122N22O19
Molecular Weight:
1756.01
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.32
Recommended Products
Assay
>97% (HPLC)
form
solid
storage temp.
−20°C
Gene Information
human ... CTSD(1509) , CTSE(1510)
mouse ... CTSD(13033) , CTSE(13034)
rat ... CTSD(171293) , CTSE(25424)
Amino Acid Sequence
MCA-Gly-Lys-Pro-Ile-Leu-Phe-Phe-Arg-Leu-Lys-DNP-Arg-NH2
Application
7-methoxycoumarin-4-acetyl-Gly-Lys-Pro-Ile-Leu-Phe-Phe-Arg-Leu-Lys-(DNP)-DArg-amide has been used as a fluorogenic substrate for the quantification of cathepsin D in decapod crustaceans.
Packaging
Bottomless glass bottle. Contents are inside inserted fused cone.
Substrates
Substrate for cathepsin D and E.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Cold-adapted digestive aspartic protease of the clawed lobsters Homarus americanus and Homarus gammarus: biochemical characterization
Liliana Rojo
Marine Biotechnology (2013)
Is digestive cathepsin D the rule in decapod crustaceans?
Martinez A D, et al.
Comparative Biochemistry and Physiology. Part B, Biochemistry & Molecular Biology, 215, 31-38 (2018)
Multiple effects of pepstatin A on Trypanosoma cruzi epimastigote forms.
Sangenito LS
Parasitology Research (2012)
Diana Martínez-Alarcón et al.
Comparative biochemistry and physiology. Part B, Biochemistry & molecular biology, 215, 31-38 (2017-10-17)
Cathepsin D is an aspartic endopetidase with typical characteristics of lysosomal enzymes. Cathepsin D activity has been reported in the gastric fluid of clawed lobsters where it acts as an extracellular digestive enzyme. Here we investigate whether cathepsin D is
Bianca Santos Henriques et al.
Frontiers in physiology, 11, 509310-509310 (2021-02-02)
Rhodnius prolixus is one important vector for the parasite Trypanosoma cruzi in Latin America, where Chagas disease is a significant health issue. Although R. prolixus is a model for investigations of vector-parasite interaction and transmission, not much has been done
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service