Skip to Content
Merck
All Photos(1)

Key Documents

L0375

Sigma-Aldrich

LAH Peptide

powder, mixture of soluble peptides and amino acids

Synonym(s):

LAH

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.85

product name

Lactalbumin enzymatic hydrolysate, a mixture of soluble peptides and amino acids

grade

for molecular biology

sterility

non-sterile

form

powder

storage temp.

2-8°C

suitability

nonselective for Escherichia coli
nonselective for coliforms

Looking for similar products? Visit Product Comparison Guide

General description

Lactalbumin enzymatic hydrolysate (LAH) is derived from the enzymatic digest of lactalbumin, often used in culture media. LAH contains a mixture of highly-soluble peptides and amino acids.

Application

Suitable for use as nutritional source in fermentations and microbial media. It is also suitable as a supplement for eukaryotic cell culture medium.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service