Key Documents
HS234634
Pipette Tip Disposal Box
clear (disposal box)
About This Item
UNSPSC Code:
41122800
NACRES:
NB.01
material
acrylic
clear (disposal box)
feature
Pipette tips
packaging
pack of 1 ea
manufacturer/tradename
Heathrow Scientific HS234634
L × W × H
154 mm × 171 mm × 206 mm
General description
  • The 3/8 in. (10 mm) thick acrylic provides protection from Beta radiation
  • The large, hinged lid allows easy disposal of contents
  • Use in conjunction with small biohazard disposal bags, if hazardous fluids are not acrylic friendly
The Pipette Tip Disposal Box′s quick flip of port cover makes tip ejection easy. It is convenient for one-handed disposal of used pipette tips and eliminates any need to handle pipette tips used with hazardous. fluids.