Skip to Content
Merck
All Photos(2)

Key Documents

HPA047533

Sigma-Aldrich

Anti-SEC11A antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody

Synonym(s):

SEC11L1, SPC18, SPCS4A, sid2895

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL

immunogen sequence

SPIVVVLSGSMEPAFHRGDLLFLTN

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SEC11A(23478)

General description

SEC11A (SEC11 homolog A, signal peptidase complex subunit) codes for the SPC18 (signal peptidase complex 18) protein. SPC18 is a subunit of the signal peptidase complex (SPC). It is located on human chromosome 15q25.2.

Immunogen

SEC11 homolog A, signal peptidase complex subunit

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

SPC18 (signal peptidase complex 18) plays a major role in the endoplasmic reticulum‐Golgi secretory pathway. It controls the secretion of several secretory proteins. SPC18 also modulates the phosphorylation of epidermal growth factor receptor (EGFR) and regulates downstream targets.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST90447

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Clinicopathological significance of SPC 18 in colorectal cancer: SPC 18 participates in tumor progression
Hattori T, et al.
Cancer Science, 108(1), 143-150 (2017)
Yuji Yamamoto et al.
Pathobiology : journal of immunopathology, molecular and cellular biology, 87(4), 254-261 (2020-06-22)
Esophageal cancer is the sixth most common malignancy worldwide. Signal peptidase complex 18 (SPC18) protein, which is encoded by the SEC11A gene, is one of the subunits of the signal peptidase complex and plays an important role in the secretion
Acquired genomic copy number changes in CML patients with the Philadelphia chromosome (Ph+)
Lu X, et al.
Cancer Genetics, 205(10), 513-518 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service