Skip to Content
Merck
All Photos(7)

Key Documents

HPA044653

Sigma-Aldrich

Anti-SPI1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-OF, Anti-PU.1, Anti-SFPI1, Anti-SPI-1, Anti-SPI-A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

EDLVPYDTDLYQRQTHEYYPYLSSDGESHSDHYWDFHPHHVHSEFESFAENNFTELQSVQPPQLQQLYRHMELEQMHVLDTPMVPPHPSLGHQVSYLPRMCLQYPSLSPAQPSSDEEEG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SPI1(6688)

General description

Spi-1 proto-oncogene (SPI1), also called PU.1, belongs to E26 transformation-specific (ETS) transcription factor family and is highly expressed in hematopoietic stem cells. SPI1 gene is mapped to human chromosome 11p11.2. It has N-terminal transcriptional activation domain with acidic amino acid sub domains, a central proline, glutamic acid, serine, and threonine-rich (PEST) region and a C-terminal ETS domain.

Immunogen

Spi-1 proto-oncogene

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Spi-1 proto-oncogene (SPI1) mediates transcriptional regulation by DNA binding and protein interactions via its ETS domain. SPI1 is crucial for the fetal lymphoid and myeloid differentiation. PU.1 interaction with GFI-1 (growth factor independence-1) is essential for macrophage granulocyte development. High expression of Spi1 leads to senescence in erythroid progenitors and is regarded as critical step to target leukemia. It also interacts with microRNA, miR-424 and modulates macrophage differentiation. Deletions in the SPI1 gene is implicated in acute myeloid leukemia (AML). SPI1 gene inactivation results in fetal death.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST84738

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Senescence is a Spi1/PU. 1-induced anti-proliferative mechanism in primary hematopoietic cells
Delestre L, et al.
Haematologica, 102(11), 1850?1860-1850?1860 (2017)
The interplay between the master transcription factor PU. 1 and miR-424 regulates human monocyte/macrophage differentiation
Rosa A, et al.
Proceedings of the National Academy of Sciences of the USA, 104(50), 19849-19854 (2007)
Inactivation of PU. 1 in adult mice leads to the development of myeloid leukemia
Metcalf D, et al.
Proceedings of the National Academy of Sciences of the USA, 103(5), 1486-1491 (2006)
The transcriptional repressor GFI-1 antagonizes PU. 1 activity through protein-protein interaction
Dahl R, et al.
The Journal of Biological Chemistry, 282(9), 6473-6483 (2007)
Heterozygous deletion of the PU. 1 locus in human AML
Bonadies N, et al.
Blood, 115(2), 331-334 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service