Skip to Content
Merck
All Photos(2)

Key Documents

HPA042257

Sigma-Aldrich

Anti-RP1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

MAPRE2, Microtubule-associated protein RP/EB family member 2, Anti-Dcdc4a, Anti-Retinitis pigmentosa 1 (autosomal dominant)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:500-1:1000

immunogen sequence

VTCSPCEMCTVNKAYSPKETCNPSDTFFPSDGYGVDQTSMNKACFLGEVCSLTDTVFSDKACAQKENHTYEGACPIDETYVPVNVCNTIDFLNSKENTYTDNLDSTEELERGDDIQKDLNILTDPEYKNGFNTLVSHQNVSNLSSCG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RP1(6101)

General description

Retinitis pigmentosa-1 (RP1) is a photoreceptor microtubule-associated protein. RP1 belongs to end binding protein 1(EB1) protein family, which interacts with adenomatous polyposis coli (APC). RP1 is located on human chromosome 8.

Immunogen

retinitis pigmentosa 1 (autosomal dominant) recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Retinitis pigmentosa-1 (RP1) regulates microtubule formation. Casein kinase II (CK2) a protein kinase when phosphorylates RP1 at Ser(236) aids cell adhesion.RP1 contributes to maintain the architecture of photoreceptor outer segments. Dysfunctioning of RP1 leads to neurodegenerative diseases. Mutations in RP1 gene is known to cause retinitis pigmentosa. Certain regions of RP1 may also cause autosomal recessive rod-cone dystrophy.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71374

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Samuel McLenachan et al.
Experimental eye research, 218, 109024-109024 (2022-03-11)
Type 2 idiopathic macular telangiectasia (MacTel-2) is a progressive adult-onset macular disease associated with bilateral perifoveal vascular changes, Muller cell degeneration and increased blood-retinal barrier permeability. The pathophysiological mechanisms of MacTel-2 remain unclear, however it was previously reported that anti-retinal
Linkage mapping of autosomal dominant retinitis pigmentosa (RP1) to the pericentric region of human chromosome 8
Blanton,, et al.
Genomics, 11(4), 857-869 (1991)
RP1 is a phosphorylation target of CK2 and is involved in cell adhesion
Stenner,, et al.
PLoS ONE, 8(7), e67595-e67595 (2013)
The retinitis pigmentosa 1 protein is a photoreceptor microtubule-associated protein
Liu, Qin, et al.
The Journal of Neuroscience, 24(29), 6427-6436 (2004)
Molecular basis for photoreceptor outer segment architecture
Goldberg,, et al.
Progress in Retinal and Eye Research, 55, 52-81 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service