Skip to Content
Merck
All Photos(4)

Key Documents

HPA040670

Sigma-Aldrich

Anti-FOXC1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-ARA, Anti-FKHL7, Anti-FREAC3, Anti-Forkhead box C1, Anti-IGDA, Anti-IHG1, Anti-IRID1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

YPGQQQNFHSVREMFESQRIGLNNSPVNGNSSCQMAFPSSQSLYRTSGAFVYDCSKF

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FOXC1(2296)

General description

Forkhead box C1 (FOXC1) is encoded by the gene mapped to human chromosome 6p25.3. The gene is widely expressed in mesenchyme and codes for a member of the FOX family, which is characterized by a distinct DNA-binding forkhead domain.

Immunogen

forkhead box C1 recombinant protein epitope signature tag (PrEST)

Application

Anti-FOXC1 antibody produced in rabbit has been used in immunoprecipitation assay.

Biochem/physiol Actions

Forkhead box C1 (FOXC1) is involved in various developmental processes such as somatic, cardiovascular, kidney, eye, skull and cortical development. Mutation in the gene leads to Dandy-Walker malformation (DWM), Axenfeld-Rieger syndrome (ARS), congenital glaucoma and cerebral small-vessel disease (CSVD). Increased expression of FOXC1 improves invasion capacity of basal-like breast cancer (BLBC) in vitro and stimulates cancer development and metastasis in vivo.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST81794

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kimberly A Aldinger et al.
Nature genetics, 41(9), 1037-1042 (2009-08-12)
Dandy-Walker malformation (DWM), the most common human cerebellar malformation, has only one characterized associated locus. Here we characterize a second DWM-linked locus on 6p25.3, showing that deletions or duplications encompassing FOXC1 are associated with cerebellar and posterior fossa malformations including
Curtis R French et al.
The Journal of clinical investigation, 124(11), 4877-4881 (2014-09-25)
Patients with cerebral small-vessel disease (CSVD) exhibit perturbed end-artery function and have an increased risk for stroke and age-related cognitive decline. Here, we used targeted genome-wide association (GWA) analysis and defined a CSVD locus adjacent to the forkhead transcription factor
Hou Dong Zuo et al.
Journal of B.U.ON. : official journal of the Balkan Union of Oncology, 21(4), 818-825 (2016-09-30)
To investigate the role of high forkhead box C1 (FOXC1) expression in basal-like breast cancer (BLBC) in vitro and vivo and the underlying regulatory mechanism. The lentivirus vector with green fluorescent protein (GFP) was used. MDA-MB-231 cells expressing consistently high
David J DeGraff et al.
Laboratory investigation; a journal of technical methods and pathology, 94(7), 726-739 (2014-05-21)
The forkhead box (Fox) superfamily of transcription factors has essential roles in organogenesis and tissue differentiation. Foxa1 and Foxa2 are expressed during prostate budding and ductal morphogenesis, whereas Foxa1 expression is retained in adult prostate epithelium. Previous characterization of prostatic
Morteza Seifi et al.
Human mutation, 38(2), 169-179 (2016-11-03)
Mutations in the forkhead box C1 gene (FOXC1) cause Axenfeld-Rieger syndrome (ARS). Here, we investigated the effect of four ARS missense variants on FOXC1 structure and function, and examined the predictive value of four in silico programs for all 31

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service