Skip to Content
Merck
All Photos(7)

Documents

HPA023280

Sigma-Aldrich

Anti-CNP antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab3

Synonym(s):

Anti-2',3'-cyclic-nucleotide 3'-phosphodiesterase, Anti-CNP, Anti-CNPase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:1000-1:2500

immunogen sequence

PGVLHCTTKFCDYGKAPGAEEYAQQDVLKKSYSKAFTLTISALFVTPKTTGARVELSEQQLQLWPSDVDKLSPTDNLPRGSRAHITL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CNP(1267)

General description

CNP (2′,3′-cyclic nucleotide 3′ phosphodiesterase) is highly expressed in brain and is used as a marker for oligodendrocytes. It is a member of the 2H phosphoesterase superfamily. The gene is mapped to human chromosome 17q.

Immunogen

2′,3′-cyclic-nucleotide 3′-phosphodiesterase recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

CNP (2′,3′-cyclic nucleotide 3′ phosphodiesterase) is responsible for the conversion of 2′,3′-cAMP (cyclic adenosine monophosphate) to 2′-AMP. It also plays a crucial role in the renal 2′,3′-cAMP-adenosine pathway. It is needed for the outgrowth in oligodendrocytes and association with microtubules, cytoskeleton, and RNA. Upon aging, low expression of CNP is associated with a catatonia-depression syndrome in humans. CNP also participates in the interferon-mediated suppression of HIV (human immunodeficiency virus) and HBV (hepatitis B virus) production.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75816

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yanpan Gao et al.
Brain pathology (Zurich, Switzerland), 32(4), e13047-e13047 (2022-01-12)
The hippocampus and entorhinal cortex (EC), the earliest affected areas, are considered relative to early memory loss in Alzheimer's disease (AD). The hippocampus is composed of heterogeneous subfields that are affected in a different order and varying degrees during AD
T J Sprinkle et al.
Genomics, 16(2), 542-545 (1993-05-01)
The human 2',3'-cyclic nucleotide 3'-phosphohydrolase (CNP) gene is located on chromosome 17, as determined by PCR of somatic cell hybrid DNA panels and confirmed using a mouse-human hybrid containing only human chromosome 17. A polymorphic site (C, T) was previously
Hui Ma et al.
PloS one, 8(11), e80769-e80769 (2013-11-22)
2',3'-cyclic nucleotide 3'-phosphodiesterase (CNP) is a member of the interferon-stimulated genes, which includes isoforms CNP1 and CNP2. CNP1 is locally expressed in the myelin sheath but CNP2 is additionally expressed at low levels outside the nervous system. CNPs regulate multiple
Nora Hagemeyer et al.
EMBO molecular medicine, 4(6), 528-539 (2012-04-05)
Severe mental illnesses have been linked to white matter abnormalities, documented by postmortem studies. However, cause and effect have remained difficult to distinguish. CNP (2',3'-cyclic nucleotide 3'-phosphodiesterase) is among the oligodendrocyte/myelin-associated genes most robustly reduced on mRNA and protein level
Edwin K Jackson et al.
American journal of physiology. Renal physiology, 307(1), F14-F24 (2014-05-09)
Energy depletion increases the renal production of 2',3'-cAMP (a positional isomer of 3',5'-cAMP that opens mitochondrial permeability transition pores) and 2',3'-cAMP is converted to 2'-AMP and 3'-AMP, which in turn are metabolized to adenosine. Because the enzymes involved in this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service