Skip to Content
Merck
All Photos(8)

Key Documents

HPA021146

Sigma-Aldrich

Anti-ATXN2 antibody produced in rabbit

enhanced validation

affinity isolated antibody, buffered aqueous glycerol solution, Ab3

Synonym(s):

Anti-Ataxin-2, Anti-Spinocerebellar ataxia type 2 protein, Anti-Trinucleotide repeat-containing gene 13 protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
conjugate:
unconjugated
application:
IHC
clone:
polyclonal
species reactivity:
human
citations:
9
technique(s):
immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:500-1:1000

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

RHPRNHRVSAGRGSISSGLEFVSHNPPSEAATPPVARTSPSGGTWSSVVSGVPRLSPKTHRPRSPRQNSIGNTPSGPVLASPQAGIIPTEAVAMPIPAASPTPASPASNRAVTPSSEAKDSRLQDQRQNSPAGNKENIKPNET

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ATXN2(6311)

General description

The gene ataxin-2 (ATXN2) is mapped to human chromosome 12q24.1. The protein belongs to ataxin-2 family. ATXN2 is present at the endoplasmic reticulum, plasma membrane and trans-Golgi network depending on the interacting partner.

Immunogen

Ataxin-2 recombinant protein epitope signature tag (PrEST)

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunofluorescence (1 paper)

Biochem/physiol Actions

Unstable expansion of ataxin-2 (ATXN2) polyglutamine domain causes neurodegenerative diseases including spinocerebellar ataxia type-2, frontotemporal dementia and amyotrophic lateral sclerosis. Mode of action involves ATXN2 association with polyribosomes and poly(A) binding protein, thereby interfering with the translational regulation of particular mRNAs. Single nucleotide polymorphism in ATXN2 is associated with schizophrenia. ATXN2 is associated with the endocytosis complex and plays a role in endocytic receptor cycling.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73803

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexander S Brown et al.
Proceedings of the National Academy of Sciences of the United States of America, 115(52), E12407-E12416 (2018-12-12)
The genetically heterogeneous spinocerebellar ataxias (SCAs) are caused by Purkinje neuron dysfunction and degeneration, but their underlying pathological mechanisms remain elusive. The Src family of nonreceptor tyrosine kinases (SFK) are essential for nervous system homeostasis and are increasingly implicated in
Adriano Chiò et al.
Neurology, 84(3), 251-258 (2014-12-21)
To analyze the frequency and clinical characteristics of patients with amyotrophic lateral sclerosis (ALS) with intermediate-length (CAG) expansion (encoding 27-33 glutamines, polyQ) in the ATXN2 gene, in a population-based cohort of Italian patients with ALS (discovery cohort), and to replicate
Serena Lattante et al.
Neurology, 83(11), 990-995 (2014-08-08)
The aim of this study was to establish the frequency of ATXN2 polyglutamine (polyQ) expansion in large cohorts of patients with amyotrophic lateral sclerosis (ALS), frontotemporal dementia (FTD), and progressive supranuclear palsy (PSP), and to evaluate whether ATXN2 could act
S Sahba et al.
Genomics, 47(3), 359-364 (1998-04-16)
Spinocerebellar ataxia type 2 (SCA2) is a member of a group of neurodegenerative diseases that are caused by instability of a DNA CAG repeat. We report the genomic structure of the SCA2 gene. Its 25 exons, encompassing approximately 130 kb
Fuquan Zhang et al.
Neuroscience letters, 562, 24-27 (2013-12-18)
Schizophrenia (SZ) is a severe mental disorder characterized by multiple neurodevelopmental dysfunctions including a breakdown of thinking process and a deficit of typical emotional responses. Ataxin-2 (ATXN2) plays vital roles in cell proliferation and growth, and functional mutations of ATXN2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service