Skip to Content
Merck
All Photos(3)

Key Documents

HPA020884

Sigma-Aldrich

Anti-SOCS5 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CIS-6, Anti-Cytokine-inducible SH2 protein 6, Anti-Cytokine-inducible SH2-containing protein 5, Anti-SOCS-5, Anti-Suppressor of cytokine signaling 5

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

HLIKQHTAPVSPHSTFFDTFDPSLVSTEDEEDRLRERRRLSIEEGVDPPPNAQIHTFEATAQVNPLYKLGPKLAPGMTEISGDSSAIPQANCDSEEDTTTLCLQSRRQKQRQISGDSHTHVSRQGAWKVHTQI

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SOCS5(9655)

General description

The gene SOCS5 (suppressor of cytokine signaling 5) is mapped to human chromosome 2p21. It belongs to SOCS family of proteins. The protein contains a Src homology-2 and SOCS box domains.

Immunogen

Suppressor of cytokine signaling 5 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

SOCSs (suppressor of cytokine signaling) are known as negative regulators of cytokine signaling. Presence of SOCS5 results in degradation of EGFR (epidermal growth factor receptor) and inhibition of JAK1 (janus kinase 1) kinase activity. It suppresses IL (interleukin)-4, IL-6 and leukemia inhibitory factor-mediated signaling. Single nucleotide polymorphism in SOCS5 is linked with bronchial asthma.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86447

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adam Weizman et al.
Inflammatory bowel diseases, 20(3), 525-533 (2014-02-04)
Pyoderma gangrenosum (PG) and erythema nodosum (EN) are the most common cutaneous manifestations of inflammatory bowel disease (IBD) but little is known regarding their etiopathogenesis. We performed a case-control study comparing characteristics between IBD patients with a documented episode of
I V Saltykova et al.
Vestnik Rossiiskoi akademii meditsinskikh nauk, (7)(7), 53-56 (2013-12-18)
To investigate the role of polymorphic variants of immune-response modifying genes in predisposition to asthma. The analysis of restriction fragments length polymorphism was used to investigate 10 single-nucleotide polymorphisms: IFNG rs2069705, IFNGR2 rs17880053, IL4 rs 2070874, IL4RA rs 1805010, GATA3
Edmond M Linossi et al.
PloS one, 8(8), e70536-e70536 (2013-08-31)
Suppressor of Cytokine Signaling (SOCS)5 is thought to act as a tumour suppressor through negative regulation of JAK/STAT and epidermal growth factor (EGF) signaling. However, the mechanism/s by which SOCS5 acts on these two distinct pathways is unclear. We show
Edith Kario et al.
The Journal of biological chemistry, 280(8), 7038-7048 (2004-12-14)
Suppressors of cytokine signaling (SOCS) are Src homology-2-containing proteins originally identified as negative regulators of cytokine signaling. Accumulating evidence indicates a role for SOCS proteins in the regulation of additional signaling pathways including receptor tyrosine kinases. Notably, SOCS36E, the Drosophila

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service