Skip to Content
Merck
All Photos(1)

Key Documents

HPA012873

Sigma-Aldrich

Anti-PDE5A antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CGB-PDE antibody produced in rabbit, Anti-cGMP-binding cGMP-specific phosphodiesterase antibody produced in rabbit, Anti-cGMP-specific 3′,5′-cyclic phosphodiesterase antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

GKKNKVIGVCQLVNKMEENTGKVKPFNRNDEQFLEAFVIFCGLGIQNTQMYEAVERAMAKQMVTLEVLSYHASAAEEETRELQSLAAAVVPSAQTLKITDFSFSDFELSDLETALCTIRMFTDLNLVQNFQMKHEVLCRWIL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PDE5A(8654)

Looking for similar products? Visit Product Comparison Guide

General description

Cyclic guanosine monophosphate (cGMP)-specific 3′, 5′-cyclic phosphodiesterase (PDE5A) is an 875-amino acid protein. It is expressed in platelets and smooth muscle cells. PDE5A is a homodimer with two regulatory domains that is, a carboxyl-terminal catalytic domain and an amino-terminal domain containing two allosteric cGMP-binding sites. The gene encoding this protein is present on chromosome 4q25-27.

Immunogen

cGMP-specific 3′,5′-cyclic phosphodiesterase recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Cyclic guanosine monophosphate (cGMP)-specific 3′, 5′-cyclic phosphodiesterase (PDE5A) is involved in the regulation of nitric-oxide-generated cGMP, smooth muscle relaxation and neuronal survival. It hydrolyzes cGMP to 5′-GMP. PDE5A is involved in pain signaling during migraine-like attacks. PDE5A has been shown to be overexpressed in squamous and transitional cell carcinomas.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86842

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dong I Lee et al.
Nature, 519(7544), 472-476 (2015-03-25)
Cyclic guanosine monophosphate (cGMP) is a second messenger molecule that transduces nitric-oxide- and natriuretic-peptide-coupled signalling, stimulating phosphorylation changes by protein kinase G. Enhancing cGMP synthesis or blocking its degradation by phosphodiesterase type 5A (PDE5A) protects against cardiovascular disease. However, cGMP
Annamaria Morelli et al.
Endocrinology, 145(5), 2253-2263 (2004-02-07)
By real-time RT-PCR and Western blot analysis, we found that phosphodiesterase type 5 (PDE5) mRNA and protein abundance was several fold higher in human male than in female reproductive tracts. The highest mRNA level (>1 x 10(7) molecules/microg total RNA)
Sergei D Rybalkin et al.
The EMBO journal, 22(3), 469-478 (2003-01-30)
cGMP-specific, cGMP-binding phosphodiesterase (PDE5) regulates such physiological processes as smooth muscle relaxation and neuronal survival. PDE5 contains two N-terminal domains (GAF A and GAF B), but the functional roles of these domains have not been determined. Here we show that
G A Piazza et al.
Cancer research, 61(10), 3961-3968 (2001-05-19)
Exisulind (Aptosyn) is a novel antineoplastic drug being developed for the prevention and treatment of precancerous and malignant diseases. In colon tumor cells, the drug induces apoptosis by a mechanism involving cyclic GMP (cGMP) phosphodiesterase inhibition, sustained elevation of cGMP
K Loughney et al.
Gene, 216(1), 139-147 (1998-08-26)
Human cGMP-binding, cGMP-specific 3',5'-cyclic nucleotide phosphodiesterase (PDE5A) cDNAs were isolated. A 3.1-kb composite DNA sequence assembled from overlapping cDNAs encodes an 875-amino-acid protein with a predicted molecular mass of 100012 Da (PDE5A1). Extracts prepared from yeast expressing human PDE5A1 hydrolyzed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service