Skip to Content
Merck
All Photos(11)

Key Documents

HPA009672

Sigma-Aldrich

Anti-SLC9A3R1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-EBP50, Anti-Ezrin-radixin-moesin-binding phosphoprotein 50, Anti-NHERF-1, Anti-Na(+)/H(+) exchange regulatory cofactor NHE-RF, Anti-Regulatory cofactor of Na(+)/H(+) exchanger, Anti-Sodium-hydrogen exchanger regulatory factor 1, Anti-Solute carrier family 9 isoform A3 regulatory

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43
conjugate:
unconjugated
application:
IHC
clone:
polyclonal
species reactivity:
human
citations:
7
technique(s):
immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
recombinant expression
independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

GENVEKETHQQVVSRIRAALNAVRLLVVDPETDEQLQKLGVQVREELLRAQEAPGQAEPPAAAEVQGAGNENEPREADKSHPEQRELRPRLCTMKKGPSGYGFNLHSD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC9A3R1(9368)

General description

SLC9A3R1 (solute carrier 9A3 receptor 1), commonly known as Na+/H+ exchanger regulatory factor 1 (NHERF1), is an ezrin-radixin-moesin (ERM)-binding phosphoprotein 50. This gene is localized to human chromosome 17q25.1, and codes for a scaffold protein. This protein consists of two tandem PDZ domains and a C-terminal ezrin-binding region.

Immunogen

Ezrin-radixin-moesin-binding phosphoprotein 50 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

SLC9A3R1 (solute carrier 9A3 receptor 1) protein plays an essential role in the binding of integral membrane proteins to cytoskeleton. Thus, it is thought to be a key player in cell signaling involved in cytoskeleton dynamics. Its expression is linked with multiple types of cancer and their aggressiveness. In breast cancer, its loss of nuclear expression is linked with decreased survival. Studies in colon adenocarcinoma HT-29 epithelial cells show that this protein plays an essential role in cellular proliferation through Gq-mediated signalling pathways. It also controls EGF (epidermal growth factor) and neurotensin signaling in HT-29 cell line. It plays an essential role in the biogenesis of microvilli, where it is critical for the retention of specific proteins in the microvilli. Activation by Rac1 protein results in conformation changes in SLC9A3R1 protein, which leads to the anchoring of cystic fibrosis transmembrane conductance regulator (CFTR) in the cell membrane.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70135

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wade A Kruger et al.
Biochemical and biophysical research communications, 432(4), 568-573 (2013-03-05)
Neurotensin receptors (NT-R) and the epidermal growth factor receptors (EGF-R) are commonly overexpressed in many epithelial origin tumours. In addition to their role as mitogenic mediators through specific cell signalling, recent studies indicate that the activity/expression of scaffold proteins responsible
Cláudia A Loureiro et al.
Science signaling, 8(377), ra48-ra48 (2015-05-21)
The peripheral protein quality control (PPQC) checkpoint removes improperly folded proteins from the plasma membrane through a mechanism involving the E3 ubiquitin ligase CHIP (carboxyl terminus of Hsc70 interacting protein). PPQC limits the efficacy of some cystic fibrosis (CF) drugs
A Paradiso et al.
Cell death & disease, 4, e904-e904 (2013-11-10)
Our purpose was to investigate whether Na(+)/H(+) exchanger regulatory factor 1 (NHERF1) expression could be linked to prognosis in invasive breast carcinomas. NHERF1, an ezrin-radixin-moesin (ERM) binding phosphoprotein 50, is involved in the linkage of integral membrane proteins to the
Qiqi Wang et al.
Cell death & disease, 9(6), 668-668 (2018-06-06)
Cervical cancer is one of the most lethal types of cancer in female. Aberrant activation of Wnt/β-catenin signaling pathway has been found to be involved in cervical cancer development and progression, whereas the underlying molecular mechanisms remain poorly understood. The
Qiqi Wang et al.
International journal of cancer, 144(7), 1619-1632 (2018-09-20)
HPV16 is the predominant type of HPV causing invasive cervical cancer. However, the underlying molecular mechanism of the unparalleled carcinogenic power of HPV16 compared to other types of high-risk (HR)-HPV including HPV18 remains elusive. The PDZ binding motif (PBM) of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service