Skip to Content
Merck
All Photos(5)

Key Documents

HPA007029

Sigma-Aldrich

Anti-COL6A2 antibody produced in rabbit

enhanced validation

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Collagen α-2(VI) chain precursor antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:50-1:200

immunogen sequence

TTERNNNCPEKTDCPIHVYFVLDTSESVTMQSPTDILLFHMKQFVPQFISQLQNEFYLDQVALSWRYGGLHFSDQVEVFSPPGSDRASFIKNLQGISSFRRGTFTDCALANMTEQIRQDR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... COL6A2(1292)

Immunogen

Collagen α-2(VI) chain precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-COL6A2 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

COL6A2 (Collagen, type VI, α2) functions as a ligand for membrane-spanning proteoglycan NG2. It directly binds with the central region of NG2 and forms the rod like structure between the N and C-terminal globules. It uses NG2 as a cell surface receptor for maintaining the pericellular matrix structure. It also connects the extracellular matrix with cells. Mutation in COL6A2 causes an autosomal dominant myopathy, Bethlem myopathy.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70481

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Tillet et al.
The Journal of biological chemistry, 272(16), 10769-10776 (1997-04-18)
NG2 is a membrane-spanning proteoglycan with a primary structure unique among cell surface or extracellular matrix proteins. To characterize the interaction between NG2 and extracellular matrix proteins, we have used a eukaryotic expression system to produce and purify several recombinant
Zhumur Ghosh et al.
Cancer research, 71(14), 5030-5039 (2011-06-08)
Pluripotent stem cells, both human embryonic stem cells (hESC) and human-induced pluripotent stem cells (hiPSC), can give rise to multiple cell types and hence have tremendous potential for regenerative therapies. However, the tumorigenic potential of these cells remains a great
A Nishiyama et al.
Molecular biology of the cell, 4(11), 1097-1108 (1993-11-01)
NG2 is a membrane-associated chondroitin sulfate proteoglycan with a core protein of 300 kD. Previously it was shown immunochemically that the core protein of NG2 can bind type VI collagen (Stallcup, W., Dahlin, K., and P. Healy. 1990. J. Cell
G J Jöbsis et al.
Nature genetics, 14(1), 113-115 (1996-09-01)
Among the diverse family of collagens, the widely expressed microfibrillar type VI collagen is believed to play a role in bridging cells with the extracellular matrix. Several observations imply substrate properties for cell attachment as well as association with major

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service