Skip to Content
Merck
All Photos(9)

Key Documents

HPA006881

Sigma-Aldrich

Anti-NDRG1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-DRG-1 antibody produced in rabbit, Anti-Differentiation-related gene 1 protein antibody produced in rabbit, Anti-N-myc downstream-regulated gene 1 protein antibody produced in rabbit, Anti-Nickel-specific induction protein Cap43 antibody produced in rabbit, Anti-Protein NDRG1 antibody produced in rabbit, Anti-RTP antibody produced in rabbit, Anti-Reducing agents and tunicamycin-responsive protein antibody produced in rabbit, Anti-Rit42 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

rat, human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

INVNPCAEGWMDWAASKISGWTQALPDMVVSHLFGKEEMQSNVEVVHTYRQHIVNDMNPGNLHLFINAYNSRRDLEIERPMPGTHTVTLQCPALLVVGDSSPAVDAVVECNSKLDPTKTTLL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NDRG1(10397)

Immunogen

Protein NDRG1 recombinant protein epitope signature tag (PrEST)

Application

Anti-NDRG1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

NDRG1 (N-myc downstream regulated 1) is a Rab4a (Ras-related protein Rab-4A) effector protein associated with cell proliferation, differentiation, and invasion. It localizes at the perinuclear recycling/sorting vesicles of Trans Golgi network. It is essentially required for the p53-dependent apoptosis. At the site of DNA damage, its elevated expression identifies the target genes required for mediating apoptosis. It also plays a major role in the recycling and stabilization of adhesion molecule E-cadherin. In endosome recycling, NDRG1 interacts with the membrane bound Rab4aGTPase. Reports show that NDRG1 acts as a biomarker in the development of Colorectal cancer (CRC).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70129

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fredrik S Skedsmo et al.
BMC veterinary research, 15(1), 121-121 (2019-04-29)
Mutations in the N-myc downstream-regulated gene 1 (NDRG1) can cause degenerative polyneuropathy in humans, dogs, and rodents. In humans, this motor and sensory neuropathy is known as Charcot-Marie-Tooth disease type 4D, and it is assumed that analogous canine diseases can
Anna S Berghoff et al.
Molecular cancer research : MCR, 19(4), 688-701 (2021-01-15)
Specific biological properties of those circulating cancer cells that are the origin of brain metastases (BM) are not well understood. Here, single circulating breast cancer cells were fate-tracked during all steps of the brain metastatic cascade in mice after intracardial
Zheng-Yan Zhang et al.
Cell proliferation, 53(7), e12853-e12853 (2020-06-17)
Hypoxia-inducible factors (HIFs) are thought to play important roles in the carcinogenesis and progression of VHL-deficient clear cell renal cell carcinoma (ccRCC). The roles of HIF-1/2α in VHL-deficient clear cell renal cell carcinoma were evaluated by bioinformatics analysis, immunohistochemistry staining
Qian Cheng et al.
British journal of cancer, 129(2), 237-248 (2023-05-11)
Portal vein tumour thrombus (PVTT) is the main pathway of HCC intrahepatic metastasis and is responsible for the poor prognosis of patients with HCC. However, the molecular mechanisms underlying PVTT vascular metastases have not been fully elucidated. NDRG1 expression was
Jehn-Chuan Lee et al.
Cancer letters, 355(2), 242-252 (2014-09-15)
Oral squamous cell carcinoma (OSCC) is the most common phenotype of oral cancer. N-myc downstream regulated gene 1 (NDRG1) is a modulator for cell proliferation, differentiation, and invasion. The role and function of NDRG1 in OSCC cells remain inconclusive. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service