Skip to Content
Merck
All Photos(9)

Key Documents

HPA006764

Sigma-Aldrich

Anti-ACAA1 antibody produced in rabbit

enhanced validation

Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-β-Ketothiolase antibody produced in rabbit, Anti-3-Ketoacyl-CoA thiolase, peroxisomal precursor antibody produced in rabbit, Anti-Acetyl-CoA acyltransferase antibody produced in rabbit, Anti-Peroxisomal 3-oxoacyl-CoA thiolase antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
conjugate:
unconjugated
application:
IF
IHC
clone:
polyclonal
species reactivity:
rat, mouse, human
citations:
4
technique(s):
immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

rat, mouse, human

enhanced validation

independent
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

GNSSQVSDGAAAILLARRSKAEELGLPILGVLRSYAVVGVPPDIMGIGPAYAIPVA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ACAA1(30)

Looking for similar products? Visit Product Comparison Guide

Immunogen

3-Ketoacyl-CoA thiolase, peroxisomal precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

ACAA1 (Acetyl-CoA acyltransferase 1) is an innate immune gene associated with the fatty acid β-oxidation in peroxisomes. It is a large molecular weight (Mr 790,000) glycoprotein present in low serum levels. It is highly involved in the endotoxin-mediated TLR4 (Toll-like receptor 4) pathway. Deficiency of ACAA1 gene causes pseudo-Zellweger syndrome. Elevated ACAA1 levels have been reported in lung and pancreatic cancerous cells. Thus, measuring ACAA1 level may add importance in the detection of specific types of cancer.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70457

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D J Culkin et al.
The Prostate, 20(2), 117-122 (1992-01-01)
Adenocarcinoma associated antigen (ACAA) is a large molecular weight protein that is normally found in low serum levels. Recent data have revealed elevations in patients with adenocarcinomas, including prostate cancer. To evaluate the relationship of ACAA levels with prostate cancer
A Bout et al.
Biochimica et biophysica acta, 1090(1), 43-51 (1991-08-27)
We have characterized the gene encoding human peroxisomal 3-oxoacyl-CoA thiolase, an enzyme operative in the peroxisomal beta-oxidation system. We found one version of this gene (gene symbol ACAA) in the human genome, in contrast to the situation in rat where
V B Pinto et al.
Cancer research, 46(12 Pt 1), 6520-6524 (1986-12-01)
The current investigation describes the purification and partial characterization of a new adenocarcinoma-associated antigen (ACAA). ACAA is a large molecular weight glycoprotein (Mr 790,000 by size chromatography on Sepharose CL-6B) that migrates in the alpha 1 region upon electrophoresis and
Joanne E Sordillo et al.
BMC medical genetics, 12, 158-158 (2011-12-14)
Polymorphisms in the endotoxin-mediated TLR4 pathway genes have been associated with asthma and atopy. We aimed to examine how genetic polymorphisms in innate immunity pathways interact with endotoxin to influence asthma risk in children. In a previous analysis of 372

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service