Skip to Content
Merck
All Photos(7)

Key Documents

HPA006456

Sigma-Aldrich

Anti-AK1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-ATP-AMP transphosphorylase antibody produced in rabbit, Anti-Adenylate kinase isoenzyme 1 antibody produced in rabbit, Anti-Myokinase antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
conjugate:
unconjugated
application:
IF
IHC
clone:
polyclonal
species reactivity:
mouse, rat, human
citations:
5
technique(s):
immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, rat, human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

EKGQLVPLETVLDMLRDAMVAKVNTSKGFLIDGYPREVQQGEEFERRIGQPTLLLYVDAGPETMTQRLLKRGETSGRVDDNEETIKKRLETYYKATEPVIAFYEKRGIVRKVNAEGS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AK1(203)

General description

AK1 (adenylate kinase 1) gene is localized to human chromosome 9q34.1. The encoded protein is composed of 194 amino acids, and is made of a single polypeptide chain. Its C-terminal contains a lysine and its N-terminal contains an acetylmethionine. It is one of the three and the most common isoforms of AK enzyme. It is a polymorphic gene, and this is attributable to its five alleles.

Immunogen

Adenylate kinase isoenzyme 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

AK1 (adenylate kinase 1) gene locus is linked to Nail patella syndrome (NPS), and this was the first autosomal linkage to be established to a genetic disorder, in humans. Mutations in this gene lead to severe AK1 deficiency in red blood cells (RBCs), and chronic nonspherocytic hemolytic anemia (CNSHA). The balanced development of fetus and placenta combined is determined by the activities of different AK1 isoforms, as this is responsible for the balance among ATP, ADP, AMP and adenosine.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST85226

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H C Duba et al.
European journal of human genetics : EJHG, 6(1), 75-79 (1998-10-22)
Nail patella syndrome (NPS) is an autosomal dominant disorder characterized by dysplasia of the nails and patella, decreased mobility of the elbow, iliac horns and in some cases nephropathy. Linkage studies have localized the NPS locus to chromosome 9q34 within
Gloria-Bottini Fulvia et al.
European journal of obstetrics, gynecology, and reproductive biology, 159(2), 273-275 (2011-08-13)
Recently our group has found that the correlation between birth weight and placental weight - an index of a balanced feto-placental unit development - is influenced by genetic factors. Since adenylate kinase locus 1 (AK₁) is a polymorphic enzyme that
Qiang Xie et al.
The Journal of reproduction and development (2024-07-16)
Calcium ions (Ca2+) play crucial roles in sperm motility and fertilization. The copine (CPNE) family comprises several Ca2+-dependent phospholipid-binding proteins. Of these, CPNE1 is extensively expressed in mammalian tissues; however, its precise role in testicular development and spermatogenesis is yet
I Von Zabern et al.
European journal of biochemistry, 68(1), 281-290 (1976-09-01)
1. Human adenylate kinase (isoenzyme AK-1-1) from skeletal muscle is a single polypeptide chain of 194 amino-acid residues with an acetylmethionine at the N-terminus and a lysine at the C-terminus. 2. The primary structure of the enzyme was determined: Ac-Met-Glu-Glu-Lys-Leu-Lys-Lys-Thr-Lys-Ile-Ile-Phe-Val-Val-Gly-Gly-Pro-Gly-Ser-Gly-Lys-Gly-Thr-Gln-Cys-Glu-Lys-Ile-Val-Gln-Lys-Tyr-Gly-Tyr-Thr-His-Leu-Ser-Thr-Gly-Asp-Leu-Leu-Arg-Ser-Glu-Val-Ser-Ser-Gly-Ser-Ala-Arg-Gly-Lys-Lys-Leu-Ser-Glu-Ile-Met-Glu-Lys-Gly-Gln-Leu-Val-Pro-Leu-Glu-Thr-Val-Leu-Asp-Met-Leu-Arg-Asp-Ala-Met-Val-Ala-Lys-Val-Asn-Thr-Ser-Lys-Gly-Phe-Leu-Ile-Asp-Gly-Tyr-Pro-Arg-Glu-Val-Gln-Gln-Gly-Glu-Glu-Phe-Glu-Arg-Arg-Ile-Gly-Gln-Pro-Thr-Leu-Leu-Leu-Tyr-Val-Asp-Ala-Gly-Pro-Glu-Thr-Met-Thr-Arg-Arg-Leu-Leu-Lys-Arg-Gly-Glu-Thr-Ser-Gly-Arg-Val-Asp-Asn-Glu-Glu-Thr-Ile-Lys-Lys-Arg-Leu-Glu-Thr-Tyr-Tyr-Lys-Ala-Thr-Glu-Pro-Val-Ile-Ala-Phe-Tyr-Glu-Lys-Arg-Gly-Ile-Val-Arg-Lys-Val-Asn-Ala-Glu-Gly-Ser-Val-Asp-Glu-Val-Phe-Ser-Gln-Val-Cys-Thr-His-Leu-Asp-Ala-Leu-Lys.
Joan-Lluis Vives Corrons et al.
Blood, 102(1), 353-356 (2003-03-22)
We report here 2 patients with chronic nonspherocytic hemolytic anemia (CNSHA) and severe red blood cell (RBC) adenylate kinase (AK) deficiency. One of these patients, a boy of Spanish origin, exhibited a neonatal icterus and splenomegaly and required blood transfusions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service