Skip to Content
Merck
All Photos(9)

Key Documents

HPA003980

Sigma-Aldrich

Anti-IL18 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-IFN-gamma-inducing factor, Anti-IL-1 gamma, Anti-IL-18, Anti-Iboctadekin, Anti-Interferon-gamma-inducing factor, Anti-Interleukin-1 gamma, Anti-Interleukin-18 precursor

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

MAAEPVEDNCINFVAMKFIDNTLYFIAEDDENLESDYFGKLESKLSVIRNLNDQVLFIDQGNRPLFEDMTDSDCRDNAPRTIFIISMYKDSQPRGMAVTISVKCEKISTLSCENKIISFKEMNPPDNIKDTKSDIIFFQRSVPGHDNKMQF

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IL18(3606)

General description

Interleukin-18 (IL18) is a cofactor that belongs to the IL-1 family. It is expressed in many mammalian cells/tissues like liver, adipose tissue and skeletal muscle. This gene is located on human chromosome 11q23.

Immunogen

interleukin 18

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Interleukin-18 (IL18) along with IL-12, induces the production of interferon γ. It participates in the modulation of lipid and glucose metabolism. It plays a major role in inflammation. Overexpression of IL-18 results in inflammatory diseases such as inflammatory myopathies and COPD (chronic obstructive pulmonary disease).
Interleukin-18 (IL18, also known as interferon-γ inducing factor) is a protein encoded by the IL18 gene in humans. The protein encoded by this gene is a pro-inflammatory cytokine and is implicated in the pathogenesis of a number of inflammatory diseases. It is a immunomodulatory cytokine that plays an important role in cellular functions against tumor development and progression. It is a key cytokine responsible for immune response and involved in the process of cancer development.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86669

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emmanuel Briend et al.
Respiratory research, 18(1), 159-159 (2017-08-24)
Increased interferon gamma (IFNγ) release occurs in Chronic Obstructive Pulmonary Disease (COPD) lungs. IFNγ supports optimal viral clearance, but if dysregulated could increase lung tissue destruction. The present study investigates which mediators most closely correlate with IFNγ in sputum in
Prithvi Kumar Singh et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 35(12), 12275-12284 (2014-11-16)
Interleukin-18 (IL-18) is one of the immunomodulatory cytokines that plays an important role in cellular functions against tumor development and progression. IL-18 (-607) C/A and (-0137) G/C gene promoter polymorphisms and their haplotypes variants are associated with risk of various
L K d C Back et al.
Tissue antigens, 84(2), 229-233 (2014-06-06)
Interleukin-18 (IL-18) is a key cytokine responsible for immune response and involved in the process of cancer development. In this case-control study, we tested whether IL-18 promoter polymorphism contributes to breast cancer susceptibility in Brazilian patients. The two groups studied
Charlotte Domblides et al.
The Journal of clinical investigation, 134(11) (2024-04-23)
The immune system can control cancer progression. However, even though some innate immune sensors of cellular stress are expressed intrinsically in epithelial cells, their potential role in cancer aggressiveness and subsequent overall survival in humans is mainly unknown. Here, we
Julie Barsalou et al.
Frontiers in immunology, 9, 2162-2162 (2018-10-16)
Gain of function (GOF) mutations affecting the inflammasome component NLRC4 are known to cause early-onset macrophage activation syndrome (MAS) and neonatal enterocolitis. Here we report a patient with a NLRC4 GOF mutation presenting with neonatal MAS efficiently treated with a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service