Skip to Content
Merck
All Photos(8)

Key Documents

HPA002038

Sigma-Aldrich

Anti-IRF4 antibody produced in rabbit

enhanced validation

Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-IRF-4 antibody produced in rabbit, Anti-Interferon regulatory factor 4 antibody produced in rabbit, Anti-LSIRF antibody produced in rabbit, Anti-Lymphocyte-specific interferon regulatory factor antibody produced in rabbit, Anti-Multiple myeloma oncogene 1 antibody produced in rabbit, Anti-NF-EM5 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43
conjugate:
unconjugated
application:
IHC
clone:
polyclonal
species reactivity:
human
citations:
5
technique(s):
immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
recombinant expression
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

SNDFEELVERSQLDISDPYKVYRIVPEGAKKGAKQLTLEDPQMSMSHPYTMTTPYPSLPAQQVHNYMMPPLDRSWRDYVPDQPHPEIPYQCPMTFGPRGHHWQGPACENGCQVTGTFYACAPPE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IRF4(3662)

Looking for similar products? Visit Product Comparison Guide

General description

Interferon regulatory factor-4 (IRF4), a transcription factor, is essential for Th (T helper)-2 effector cell differentiation. It belongs to the interferon (IFN) regulatory factor (IRF) transcription factor family. It is expressed in human lymphocytes and macrophages.

Immunogen

Interferon regulatory factor 4 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

IRF4 (interferon regulatory factor 4) acts as a transcriptional activator as well as repressor by binding to distinct DNA-binding motifs and through interaction with various additional transcription factors, most notably with the ETS (E26 transformation-specific) family member. Majorly, it contributes in the generation of plasma cells. In deficiency of IRF4, B cells show impaired expression of activation-induced deaminase and lacks class-switch recombination.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86218

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Marecki et al.
Journal of immunology (Baltimore, Md. : 1950), 163(5), 2713-2722 (1999-08-24)
IFN regulatory factor 4 (IRF4) and IFN consensus sequence binding protein (ICSBP) are highly homologous members of the growing family of IRF proteins. ICSBP expression is restricted to lymphoid and myeloid cells, whereas IRF4 expression has been reported to be
Sara Canovas Nunes et al.
Blood cancer journal, 8(2), 20-20 (2018-02-15)
Multiple myeloma is a post-germinal center B-cell neoplasm, characterized by the proliferation of malignant bone marrow plasma cells, whose survival and proliferation is sustained by growth factors and cytokines present in the bone marrow microenvironment. Among them, IL-6 triggers the
Ulf Klein et al.
Nature immunology, 7(7), 773-782 (2006-06-13)
B cells producing high-affinity antibodies are destined to differentiate into memory B cells and plasma cells, but the mechanisms leading to those differentiation pathways are mostly unknown. Here we report that the transcription factor IRF4 is required for the generation
Sylvia Marecki et al.
Journal of interferon & cytokine research : the official journal of the International Society for Interferon and Cytokine Research, 22(1), 121-133 (2002-02-16)
Gene expression is a tightly regulated process involving multiple levels of control spanning histone acetylation to protein turnover. One of the first events in this cascade is transcription, which itself is a multistep process involving protein-protein interaction and macromolecular assembly.
Ye Zheng et al.
Nature, 458(7236), 351-356 (2009-02-03)
In the course of infection or autoimmunity, particular transcription factors orchestrate the differentiation of T(H)1, T(H)2 or T(H)17 effector cells, the responses of which are limited by a distinct lineage of suppressive regulatory T cells (T(reg)). T(reg) cell differentiation and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service