Skip to Content
Merck
All Photos(7)

Key Documents

HPA001813

Sigma-Aldrich

Anti-TJP2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Tjp2 Antibody, Tjp2 Antibody - Anti-TJP2 antibody produced in rabbit, Anti-Tight junction protein 2 antibody produced in rabbit, Anti-Tight junction protein ZO-2 antibody produced in rabbit, Anti-Zona occludens 2 protein antibody produced in rabbit, Anti-Zonula occludens 2 protein antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

IGVLLMKSRANEEYGLRLGSQIFVKEMTRTGLATKDGNLHEGDIILKINGTVTENMSLTDARKLIEKSRGKLQLVVLRDSQQTLINIPSLNDSDSEIEDISEIESNRSFSPEERRHQYSDYDYHSSSEKLKERPSSREDTPSRLSRMG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TJP2(9414)

General description

Tight junction protein 2 (TJP2), a junction-associated protein, belongs to the membrane-associated guanylate kinase (MAGUK) family with two isoforms ZO (zonula occludens protein)-2A and ZO-2C. It has molecular mass of 160kDa. It is localized at the tight junctions (TJs) and adherens junctions (AJs) in epithelial and nonepithelial cells. It is present at AJs in nonepithelial cells, such as fibroblasts and cardiac muscle cells lacking TJs. In the amino-terminal region, it contains three PDZ (PSD95, Dlg1, zo-1) domains, one SH3 (src homology 3), and one GUK (guanylate kinase) domain followed by a short carboxy-terminal with non-dlg-like domain.

Immunogen

Tight junction protein ZO-2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

With PDZ domains tight junction protein 2 (TJP2) dimerize to form oligomers that provide a structural basis for the polymerization of claudins, namely the formation of tight junctions. It is involved with the junction sorting mechanism. It also binds in cross linking manner to F-actin.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST83052

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiawen Wu et al.
The Journal of biological chemistry, 282(49), 35988-35999 (2007-09-28)
Zonula occludens proteins (ZOs), including ZO1/2/3, are tight junction-associated proteins. Each of them contains three PDZ domains. It has been demonstrated that ZO1 can form either homodimers or heterodimers with ZO2 or ZO3 through the second PDZ domain. However, the
A S Fanning et al.
The Journal of biological chemistry, 273(45), 29745-29753 (1998-10-29)
The tight junction protein ZO-1 belongs to a family of multidomain proteins known as the membrane-associated guanylate kinase homologs (MAGUKs). ZO-1 has been demonstrated to interact with the transmembrane protein occludin, a second tight junction-specific MAGUK, ZO-2, and F-actin, although
Jean-Yves Métais et al.
FEBS letters, 579(17), 3725-3730 (2005-06-25)
In Drosophila, the tumor suppressor Scribble is localized at the septate junctions of epithelial cells. Its mammalian homologue, hScrib, is a basolateral protein likely associated to proteins of the cell-cell junctions. We report the direct interaction between hScrib and ZO-2
A Chlenski et al.
Biochimica et biophysica acta, 1493(3), 319-324 (2000-10-06)
One of the tight junction components, zonula occludens protein 2 (ZO-2), is expressed as two isoforms, ZO-2A and ZO-2C, in normal epithelia. In pancreatic adenocarcinoma of the ductal type ZO-2A is absent, but none of the common mechanisms of gene
E S Wittchen et al.
The Journal of biological chemistry, 274(49), 35179-35185 (1999-11-27)
Defining how the molecular constituents of the tight junction interact is a prerequisite to understanding tight junction physiology. We utilized in vitro binding assays with purified recombinant proteins and immunoprecipitation analyses to define interactions between ZO-1, ZO-2, ZO-3, occludin, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service