Skip to Content
Merck
All Photos(1)

Key Documents

HPA000953

Sigma-Aldrich

Anti-MYO18B antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Myosin-XVIIIb antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL

immunogen sequence

GSDPFSWKLPSLDYERKTKVDFDDFLPAIRKPQTPTSLAGSAKGGQDGSQRSSIHFETEEANRSFLSGIKTILKKSPEPKEDPAHLSDSSSSSGSIVSFKSADSIKSRPGIPRLAGDGGERTSPERREPGTGRKDDDVASIMKKY

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MYO18B(84700)

Immunogen

Myosin-XVIIIb recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Myosin-XVIIIb is a protein encoded by the MYO18B gene in humans. It is located on chromosome 22q12.1. The gene is may function as a tumor suppressor and its inactivation is involved in lung cancer progression. It is expressed mainly in human cardiac and skeletal muscles and, at lower levels, in testis. It can be used for the treatment of locally advanced malignant pleural mesothelioma (MPM) in humans. Alterations in this gene include both epigenetic and genetic alterations and play an important role in ovarian carcinogenesis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73434

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michela Salamon et al.
Journal of molecular biology, 326(1), 137-149 (2003-01-28)
We have characterized a novel unconventional myosin heavy chain, named MYO18B, that appears to be expressed mainly in human cardiac and skeletal muscles and, at lower levels, in testis. MYO18B transcript is detected in all types of striated muscles but
Michiho Nishioka et al.
Proceedings of the National Academy of Sciences of the United States of America, 99(19), 12269-12274 (2002-09-05)
Loss of heterozygosity on chromosome 22q has been detected in approximately 60% of advanced nonsmall cell lung carcinoma (NSCLC) as well as small cell lung carcinoma (SCLC), suggesting the presence of a tumor suppressor gene on 22q that is involved
Jun Yokota et al.
Clinical & experimental metastasis, 20(3), 189-193 (2003-05-14)
It is now widely accepted that human carcinogenesis is a multi-step process and phenotypic changes during cancer progression reflect the sequential accumulation of genetic alterations in cells. Thus, in order to understand the process of acquisition of metastatic phenotypes in
Nobutaka Edakuni et al.
Oncology research, 16(5), 235-243 (2007-02-14)
Malignant pleural mesothelioma (MPM) is closely related to exposure to asbestos, and a rapid increase in the number of MPM patients is therefore estimated to occur from 2010 to 2040 in Japan. Because MPM is refractory to conventional chemotherapy and
Nozomu Yanaihara et al.
International journal of cancer, 112(1), 150-154 (2004-08-12)
Allelic imbalance on chromosome arm 22q has been detected in 50-70% of ovarian cancers, suggesting the presence of a tumor-suppressor gene on this chromosome arm that is involved in ovarian carcinogenesis. Recently, we isolated a candidate tumor-suppressor gene, MYO18B, at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service